Skip to Content
Merck
All Photos(2)

Key Documents

M4531

Sigma-Aldrich

Meclofenamic acid sodium salt

Synonym(s):

2-[(2,6-Dichloro-3-methylphenyl)amino]benzoic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C14H10Cl2NO2Na
CAS Number:
Molecular Weight:
318.13
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

Assay

≥93% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless to faintly brownish-yellow
water: 50 mg/mL, clear, colorless to faintly yellow
water: 50 mg/mL, clear, colorless to very faintly brown

SMILES string

[Na].Cc1ccc(Cl)c(Nc2ccccc2C(O)=O)c1Cl

InChI

1S/C14H11Cl2NO2.Na.H/c1-8-6-7-10(15)13(12(8)16)17-11-5-3-2-4-9(11)14(18)19;;/h2-7,17H,1H3,(H,18,19);;

InChI key

CHGIZYUDGOIAFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Meclofenamic acid consists of two aryl moieties that are twisted in perpendicular orientation, it includes N-2,6-dichloro-3-methylphenyl group and N-2-benzoic acid group. Because of the low aqueous solubility of the compound, it is commercially available as sodium salt.[1]

Application

Meclofenamic acid sodium salt has been used:
  • in the inhibition of heat-induced denaturation of albumin[2]
  • as a pharmacological agent in the preparation of superfusion solution for electrophysiology studies[3]
  • to block the gap junctions[4]

Biochem/physiol Actions

Cyclooxygenase (IC50 = 0.6 μM) and 5-lipoxygenase (IC50 = 46.3 μM) inhibitor.
Meclofenamic acid (MFA) is a nonsteroidal anti-inflammatory, analgesic and antipyretic drug. It inhibits prostaglandin biosynthesis pathway and finds its application in the treatment of postoperative and traumatic inflammation and swelling. MFA functions in opening the potassium voltage-gated channel subfamily Q member 2/member 3 (KCNQ2/Q3).[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Homeostatic plasticity shapes cell-type-specific wiring in the retina
Tien NW, et al.
Neuron, 94(3), 656-665 (2017)
Diverse inhibitory and excitatory mechanisms shape temporal tuning in transient OFF alpha ganglion cells in the rabbit retina
Murphy-Baum BL and Taylor WR
The Journal of Physiology, 596(3), 477-495 (2018)
Boctor, A.M., et al.
Prostaglandins Leukotrienes Med., 23, 229-229 (1986)
High solubility piperazine salts of the nonsteroidal anti-inflammatory drug (NSAID) meclofenamic acid
Sanphui P, et al.
Crystal Growth & Design, 12(4), 2023-2036 (2012)
Jungryul Ahn et al.
Frontiers in cellular neuroscience, 16, 889663-889663 (2022-05-24)
Retinal prostheses have shown some clinical success in patients with retinitis pigmentosa and age-related macular degeneration. However, even after the implantation of a retinal prosthesis, the patient's visual acuity is at best less than 20/420. Reduced visual acuity may be

Questions

  1. Hello, a vascular lab conducts wire myography with human (pregnant) omental resistance arteries. U46619 (cat# D8174) is used to wake up the vessels, followed by a U46619 dose-response curve to calculate an EC80. When the inhibitor meclofenamate (cat# M4531) is added, the vessels exhibit a significantly reduced constriction response to U46619, preventing the testing of a vasodilator curve (e.g. bradykinin) due to low pre-constriction.

    1 answer
    1. After adding meclofenamate, we have not specifically studied the impact on diminished U46619-induced vasoconstriction. However, based on a literature search, it is likely that meclofenamate could lead to diminished U46619-induced vasoconstriction as it is known to alter the binding of U46619 to thromboxane receptors.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service