Skip to Content
Merck
All Photos(1)

Key Documents

SML2181

Sigma-Aldrich

SA4503

≥98% (HPLC)

Synonym(s):

1-(3,4-Dimethoxyphenethyl)-4-(3-phenylpropyl)piperazine dihydrochloride, AGY 94806, AGY94806, Cutamesine, SA 4503

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32N2O2 · 2HCl
CAS Number:
Molecular Weight:
441.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

COC1=CC=C(CCN2CCN(CCCC3=CC=CC=C3)CC2)C=C1OC

InChI

1S/C23H32N2O2.2ClH/c1-26-22-11-10-21(19-23(22)27-2)12-14-25-17-15-24(16-18-25)13-6-9-20-7-4-3-5-8-20;;/h3-5,7-8,10-11,19H,6,9,12-18H2,1-2H3;2*1H

InChI key

XWOXAKBQEMQMFH-UHFFFAOYSA-N

Biochem/physiol Actions

SA4503 is a high affinity, orally active non-opioid intracellular receptor sigma-1 (σ1R) selective agonist (IC50 = 17.4 nM against 200 nM (+)-pentazocine for binding σ1R), exhibiting ~100-fold less affinity for the sigma 2 receptor subtype (σ2R) and little or no affinity toward 36 other receptors, ion channels and second messenger systems. Oral administration is reported to elicit cerebral acetylcholine (ACh) release in rat frontal cortex and hippocampus (5-20 mg/kg p.o.) and reduce immobility time in a mouse forced swimming test (Immobility time = 214.8 s without vs. 111.6 s with 1.0 mg/kg oral dosage).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yasuharu Shinoda et al.
PloS one, 11(10), e0163992-e0163992 (2016-10-16)
Cardiovascular diseases are risk factors for depression in humans. We recently proposed that σ1 receptor (σ1R) stimulation rescued cardiac hypertrophy and heart failure induced by transverse aortic constriction (TAC) in mice. Importantly, σ1R stimulation reportedly ameliorates depression-like behaviors in rodents.
Yasuharu Shinoda et al.
Journal of pharmacological sciences, 131(3), 172-183 (2016-07-21)
Haloperidol is an antipsychotic drug that inhibits the dopamine D2 receptor among others. Haloperidol also binds the sigma-1 receptor (σ1R) and inhibits it irreversibly. A serious outcome of haloperidol treatment of schizophrenia patients is death due to sudden cardiac failure.
Dennis K Miller et al.
Pharmacology, biochemistry, and behavior, 150-151, 198-206 (2016-11-17)
This study examined the effect of the N-phenylpropyl-N'-substituted piperazine ligands SA4503 (3.4-dimethoxyphenethyl), YZ-067 (4-methoxyphenethyl), YZ-185 (3-methoxyphenethyl) and Nahas-3h (4-methoxybenzyl) on methamphetamine-induced hyperactivity in mice. In a previous study in rats, SA4503 increased methamphetamine-induced hyperactivity at a lower ligand dose and
K Matsuno et al.
European journal of pharmacology, 306(1-3), 271-279 (1996-06-13)
The binding profiles of SA4503 (1-(3,4-dimethoxyphenethyl)-4-(3-phenylpropyl)piperazine dihydrochloride), a novel sigma receptor ligand, to sigma 1 and sigma 2 receptor subtypes in guinea pig and rat brain membranes were evaluated. SA4503 showed a high affinity for the sigma 1 receptor subtype
Mori Tomohisa et al.
Synapse (New York, N.Y.), 69(11), 526-532 (2015-08-04)
Previous studies have shown that ligands of the sigma-1 receptor chaperone (Sig-1R) regulate pain-related behaviors. Clinical use of chemotherapeutics is often compromised due to their adverse side effects, particularly those related to neuropathy. Previous studies have shown that repeated administration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service