Skip to Content
Merck
All Photos(1)

Key Documents

P2607

Sigma-Aldrich

PRE-084

solid

Synonym(s):

2-(4-morpholinethyl)-1-phenylcyclohexanecarboxylate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H27NO3 · HCl
Molecular Weight:
353.88
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

solubility

H2O: ≤24 mg/mL

SMILES string

Cl.O=C(OCCN1CCOCC1)C2(CCCCC2)c3ccccc3

InChI

1S/C19H27NO3.ClH/c21-18(23-16-13-20-11-14-22-15-12-20)19(9-5-2-6-10-19)17-7-3-1-4-8-17;/h1,3-4,7-8H,2,5-6,9-16H2;1H

InChI key

QUJWFJNHTBKCLU-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

PRE-084 is a potent and highly selective σ1 opioid receptor agonist. It offers neuroprotection upon embolic stroke.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sigma-1 receptor ligand PRE-084 reduced infarct volume, neurological deficits, pro-inflammatory cytokines and enhanced anti-inflammatory cytokines after embolic stroke in rats
Allahtavakoli M and Jarrott B
Brain Research Bulletin, 85(3-4), 219-224 (2011)
Judit Hodrea et al.
International journal of molecular sciences, 24(14) (2023-07-29)
Primary open-angle glaucoma remains a global issue, lacking a definitive treatment. Increased intraocular pressure (IOP) is considered the primary risk factor of the disease and it can be caused by fibrotic-like changes in the trabecular meshwork (TM) such as increased
T Maurice et al.
British journal of pharmacology, 127(2), 335-342 (1999-06-29)
1. Sigma (sigma) receptor ligands were previously reported to alleviate learning and memory impairments on several pharmacological and pathological rodent models of amnesia. Such effect was demonstrated as involving the sigma1 subtype of sigma receptor. 2. In this study, we
Yuriko Kimura et al.
PloS one, 8(10), e75760-e75760 (2013-10-12)
Sigma-1 receptor (Sig-1R) is an integral membrane protein predominantly expressed in the endoplasmic reticulum. Sig-1R demonstrates a high affinity to various synthetic compounds including well-known psychotherapeutic drugs in the central nervous system (CNS). For that, it is considered as an
Li-Li Ji et al.
Molecular medicine reports, 16(4), 4987-4993 (2017-08-10)
Accumulating evidence has demonstrated that the σ-1 receptor (σ‑1R) possesses neuroprotective effects and is a potential novel therapeutic target for certain psychiatric diseases, including post‑traumatic stress disorder (PTSD) accompanied with anxiety disorder. It has been reported that σ‑1R agonist treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service