Skip to Content
Merck
All Photos(1)

Key Documents

C5793

Sigma-Aldrich

Ceftriaxone disodium salt hemi(heptahydrate)

third-generation cephalosporin antibiotic

Synonym(s):

Ceftriaxone disodium hemiheptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N8Na2O7S3 · 3.5H2O
CAS Number:
Molecular Weight:
661.60
EC Number:
MDL number:
UNSPSC Code:
51284150
PubChem Substance ID:
NACRES:
NA.85

form

powder or crystals

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

O.O.O.O.O.O.O.[Na+].[Na+].CO\N=C(/C(=O)N[C@H]1C2SCC(CSC3=NC(=O)C(=O)N([Na])N3C)=C(N2C1=O)C([O-])=O)c4csc(N)n4.CO\N=C(/C(=O)N[C@H]5C6SCC(CSC7=NC(=O)C(=O)N([Na])N7C)=C(N6C5=O)C([O-])=O)c8csc(N)n8

InChI

1S/2C18H18N8O7S3.4Na.7H2O/c2*1-25-18(22-12(28)13(29)23-25)36-4-6-3-34-15-9(14(30)26(15)10(6)16(31)32)21-11(27)8(24-33-2)7-5-35-17(19)20-7;;;;;;;;;;;/h2*5,9,15H,3-4H2,1-2H3,(H5,19,20,21,23,27,29,31,32);;;;;7*1H2/q;;4*+1;;;;;;;/p-4/b2*24-8-;;;;;;;;;;;/t2*9-,15-;;;;;;;;;;;/m11.........../s1

InChI key

PMRZKYOXTPBIQF-MAODNAKNSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Application

Ceftriaxone was used to study drug-induced immune hemolytic anemias, prophylactic strategies to reduce neonatal encephalopathy, pentylenetetrazole-evoked convulsions, the effect of expression, binding, and inhibition of PDP1 and other penicillin-binding proteins (PBPs) on bacterial cell wall mucopeptide synthesis.

Biochem/physiol Actions

Ceftriaxone is a third-generation cephalosporin antibiotic that disrupts the synthesis of the peptidoglycan layer of bacterial cell walls. It is effective against Gram-positive and Gram-negative bacteria. Ceftriaxone is thought to increase EAAT2 pump expression in the central nervous system and to reduce glutamatergic toxicity.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beneficial Effects of Ceftriaxone Against Pentylenetetrazole-Evoked Convulsions.
Ankica V. Jelenkovic, Marina D. Jovanovic, et al.
Exp. Biol. Med, 233, 1389-1394 (2008)
Seok-Geun Lee et al.
The Journal of biological chemistry, 283(19), 13116-13123 (2008-03-11)
Glutamate is an essential neurotransmitter regulating brain functions. Excitatory amino acid transporter (EAAT)-2 is one of the major glutamate transporters primarily expressed in astroglial cells. Dysfunction of EAAT2 is implicated in acute and chronic neurological disorders, including stroke/ischemia, temporal lobe
Filippo Lagi et al.
Journal of travel medicine, 20(2), 128-130 (2013-03-08)
We present the case of two Australian tourists aged 25 and 26  years who, after immersion in a canal in Venice, developed severe leptospirosis. After a 1-week history of fever, headache, myalgia, and vomiting they developed jaundice and renal failure.
Catherine A Ison et al.
The Lancet. Infectious diseases, 13(9), 762-768 (2013-06-15)
Effective treatment of gonorrhoea is fundamental to public health control; however, the ability of Neisseria gonorrhoeae to successively develop resistance to different treatments has hampered control efforts. The extended-spectrum cephalosporins--cefixime and ceftriaxone--have been recommended in the UK for treatment of
Shanna L Ashley et al.
Science translational medicine, 12(556) (2020-08-18)
Inhaled oxygen, although commonly administered to patients with respiratory disease, causes severe lung injury in animals and is associated with poor clinical outcomes in humans. The relationship between hyperoxia, lung and gut microbiota, and lung injury is unknown. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service