Skip to Content
Merck
All Photos(1)

Key Documents

SML1016

Sigma-Aldrich

Camalexin

≥98% (HPLC)

Synonym(s):

3-(Thiazol-2-yl)-1H-indole, Camalexine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8N2S
CAS Number:
Molecular Weight:
200.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

C12=CC=CC=C1NC=C2C3=NC=CS3

InChI

1S/C11H8N2S/c1-2-4-10-8(3-1)9(7-13-10)11-12-5-6-14-11/h1-7,13H

InChI key

IYODIJVWGPRBGQ-UHFFFAOYSA-N

Application

Camalexin has been used in quantification and tolerance assays in Arabidopsis thaliana leaves.

Biochem/physiol Actions

Camalexin is a phytoalexin isolated from cruciferous plants that exhibits antibacterial, antifungal, antiproliferative and cancer chemopreventive activities.
Camalexin is a phytoalexin isolated from cruciferous plants that exhibits antibacterial, antifungal, antiproliferative and cancer chemopreventive activities. Apparently, camalexin increases the ROS levels and is more effective in aggressive prostate cancer cells that express high ROS levels. In Arabidopsis thaliana, camalexin and salicylic acid confer resistance to Phytophthora capsici.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Camalexin quantification in Arabidopsis thaliana leaves infected with Botrytis cinerea
Savatin DV, et al.
Bio-protocol, 5(2), e1379-e1379 (2015)
Siva K Malka et al.
Frontiers in plant science, 8, 2131-2131 (2018-01-10)
Glucosinolates (GLS) are a group of plant secondary metabolites mainly found in Cruciferous plants, share a core structure consisting of a β-thioglucose moiety and a sulfonated oxime, but differ by a variable side chain derived from one of the several
Yunxia He et al.
The Plant cell, 31(9), 2206-2222 (2019-06-27)
Plant defense often depends on the synthesis and targeted delivery of antimicrobial metabolites at pathogen contact sites. The pleiotropic drug resistance (PDR) transporter PENETRATION3 (PEN3)/PDR8 in Arabidopsis (Arabidopsis thaliana) has been implicated in resistance to a variety of fungal pathogens.
Md Tohidul Islam et al.
Frontiers in plant science, 11, 852-852 (2020-08-09)
Seaweed extracts are important sources of plant biostimulants that boost agricultural productivity to meet current world demand. The ability of seaweed extracts based on either of the Phaeophyceaean species Ascophyllum nodosum or Durvillaea potatorum to enhance plant growth or suppress
Mariola Piślewska-Bednarek et al.
Plant physiology, 176(1), 538-551 (2017-11-11)
Glutathione (GSH) and indole glucosinolates (IGs) exert key functions in the immune system of the model plant Arabidopsis (

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service