Skip to Content
Merck
All Photos(1)

Documents

SML2210

Sigma-Aldrich

AubipyOMe

≥97% (NMR)

Synonym(s):

(SP-4-2)-dichloro(4,4′-dimethoxy-2,2′-bipyridine-kN1,kN1′)-Gold(1+), hexafluorophosphate(1-), [Au(4,4′-dimethoxy-2,2′-bipyridine)Cl2][PF6]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12AuCl2N2O2 · PF6
CAS Number:
Molecular Weight:
629.07
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (NMR)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[Cl-][Au+3]1([N]2=C(C=C(C=C2)OC)C3=[N]1C=CC(OC)=C3)[Cl-].F[P-](F)(F)(F)(F)F

Biochem/physiol Actions

AubipyOMe is a potent inhibitor of tartrate resistant acid phosphatase (TRAP/ACP5), a metalloenzyme expressed in activated osteoclasts and in macrophages. AubipyOMe exhibited IC50 values of 1.3 μM for TRAP5a and 1.8 μM for TRAP5b and inhibited TRAP activity in murine macrophage and human lung tissue extracts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carian E Boorsma et al.
Scientific reports, 7(1), 12570-12570 (2017-10-05)
The enzyme tartrate resistant acid phosphatase (TRAP, two isoforms 5a and 5b) is highly expressed in alveolar macrophages, but its function there is unclear and potent selective inhibitors of TRAP are required to assess functional aspects of the protein. We
Shujun Yang et al.
Molecular medicine (Cambridge, Mass.), 30(1), 89-89 (2024-06-16)
Myocardial infarction (MI) leads to enhanced activity of cardiac fibroblasts (CFs) and abnormal deposition of extracellular matrix proteins, resulting in cardiac fibrosis. Tartrate-resistant acid phosphatase 5 (ACP5) has been shown to promote cell proliferation and phenotypic transition. However, it remains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service