Skip to Content
Merck
All Photos(1)

Key Documents

SML0763

Sigma-Aldrich

SC144 hydrochloride

≥98% (HPLC)

Synonym(s):

2-(7-Fluoropyrrolo[1,2-a]quinoxalin-4-yl)hydrazide-2-pyrazinecarboxylic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11FN6O · HCl
CAS Number:
Molecular Weight:
358.76
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to light brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

InChI

1S/C16H11FN6O.ClH/c17-10-3-4-13-11(8-10)20-15(14-2-1-7-23(13)14)21-22-16(24)12-9-18-5-6-19-12;/h1-9H,(H,20,21)(H,22,24);1H

InChI key

LKFGGXYXFIICED-UHFFFAOYSA-N

Application

SC144 hydrochloride has been used:
  • as a glycoprotein 130 (gp130) inhibitor to evaluate the effect of bazedoxifene on interleukin (IL-6) mediated cell viability in pancreatic cancer cells
  • as a gp130 inhibitor to study its effects on signal transducer and activator of transcription 3 (STAT3) activation in mesenchymal precursor cells (KMM) cells
  • as an interleukin 6 signal transducer (IL-6ST) inhibitor in media to test its ability to block serum-induced migration/invasion

Biochem/physiol Actions

SC144 is a potent inhibitor of glycoprotein 130 (gp130). SC144 binds to gp130 and blocks STAT3 phosphorylation, nuclear translocation and expression of STAT3 responsive genes. SC144 inhibits the growth of several tumor cell lines, independent of p53 or hormone receptor levels, and inhibits growth of ovarain tumor xenografts in mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amy C Gross et al.
JCI insight, 3(16) (2018-08-24)
Osteosarcoma (OS), a malignant tumor of bone, kills through aggressive metastatic spread almost exclusively to the lung. Mechanisms driving this tropism for lung tissue remain unknown, though likely invoke specific interactions between tumor cells and other cells within the lung
Linhua Gan et al.
Cell reports, 30(11), 3717-3728 (2020-03-19)
Understanding the mechanisms of activity-dependent gene transcription underlying adaptive behaviors is challenging at neuronal-subtype resolution. Using cell-type specific molecular analysis in agouti-related peptide (AgRP) neurons, we reveal that the profound hunger-induced transcriptional changes greatly depend on plant homeodomain finger protein
Agnieszka A Wendorff et al.
Cancer discovery, 9(3), 436-451 (2018-12-21)
The plant homeodomain 6 gene (PHF6) is frequently mutated in human T-cell acute lymphoblastic leukemia (T-ALL); however, its specific functional role in leukemia development remains to be established. Here, we show that loss of PHF6 is an early mutational event
Xiang Chen et al.
Current cancer drug targets, 19(5), 417-427 (2018-05-02)
Elevated production of the pro-inflammatory cytokine interleukin-6 (IL-6) and dysfunction of IL-6 signaling promotes tumorigenesis and are associated with poor survival outcomes in multiple cancer types. Recent studies showed that the IL-6/GP130/STAT3 signaling pathway plays a pivotal role in pancreatic
Marion Gruffaz et al.
Cancer research, 77(24), 7094-7108 (2017-10-21)
Toll-like receptors (TLR) are conserved immune sensors mediating antimicrobial and antitumoral responses, but recent evidence implicates them in promoting carcinogenesis in certain cancers. Kaposi sarcoma is caused by infection of Kaposi sarcoma-associated herpesvirus (KSHV) and is characterized by uncontrolled neoangiogenesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service