Skip to Content
Merck
All Photos(1)

Key Documents

SML2848

Sigma-Aldrich

Nintedanib

≥98% (HPLC), powder, VEGFR, PDGFR and FGFR inhibitor

Synonym(s):

(3Z)-2,3-Dihydro-3-[[[4-[methyl[2-(4-methyl-1-piperazinyl)acetyl]amino]phenyl]amino]phenylmethylene]-2-oxo-1H-indole-6-carboxylic acid methyl ester, (Z)-3-[(4-{Methyl-[2-(4-methylpiperazin-1-yl)acetyl]amino}phenylamino)methylene]-2-oxo-2,3-dihydro-1H-indole-6-carboxylic acid methyl ester, BIBF 1120, BIBF-1120, BIBF1120, Methyl (Z)-3-(((4-(N-methyl-2-(4-methylpiperazin-1-yl)acetamido)phenyl)amino)(phenyl)methylene)-2-oxoindoline-6-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H33N5O4
CAS Number:
Molecular Weight:
539.62
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Nintedanib, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=C2)/C(C1=CC=C2C(OC)=O)=C(C3=CC=CC=C3)\NC4=CC=C(C=C4)N(C(CN5CCN(CC5)C)=O)C

InChI

1S/C31H33N5O4/c1-34-15-17-36(18-16-34)20-27(37)35(2)24-12-10-23(11-13-24)32-29(21-7-5-4-6-8-21)28-25-14-9-22(31(39)40-3)19-26(25)33-30(28)38/h4-14,19,32H,15-18,20H2,1-3H3,(H,33,38)/b29-28-

InChI key

XZXHXSATPCNXJR-ZIADKAODSA-N

Biochem/physiol Actions

Nintedanib (BIBF1120) is an orally active, potent ATP-competitive inhibitor against angiokinases VEGFR-1/2/3 (IC50 = 34/21/13 nM), FGFR-1/2/3/4 (IC50 = 69/37/108/610 nM), PDGFRα/β (IC50 = 59/65 nM), as well as Flt-3, Lck, Lyn, and Src (IC50 = 26, 16, 195, 156 nM, repectively), but not 33 other kinases. Nintedanib exhibits antiangiogenic and antifibrotic efficacy in cultures and in animal models of cancers and pulmonary fibrosis in vivo.
Orally active, potent ATP-competitive VEGFR, FGFR, PDGFR, Flt3, Lck, Lyn, Src inhibitor with in vivo antiangiogenic and antifibrotic efficacy.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei-Tien Tai et al.
Journal of hepatology, 61(1), 89-97 (2014-03-25)
Nintedanib, a triple angiokinase inhibitor, is currently being evaluated against advanced HCC in phase I/II clinical trials. Here, we report the underlying molecular mechanism by which nintedanib (BIBF-1120) induces an anti-HCC effect. To further elucidate whether the effect of nintedanib
Xiaoping Cai et al.
Clinical and experimental pharmacology & physiology, 40(9), 662-670 (2013-07-04)
In the present study, we tested whether serum amyloid A (SAA) protein, an established biomarker of inflammation, also plays a role in stimulating neovascularization. To evaluate this possibility, human carotid artery endothelial (HCtAE) cells were cultured and cellular migration and
Raquel Frenedoso da Silva et al.
Cell and tissue research, 379(2), 407-420 (2019-09-02)
The antiangiogenic therapy for prostate cancer with Nintedanib, a potent inhibitor of important growth factor receptors, has been proven to delay tumor progression and arrest tumor growth; thus, the aim herein is to evaluate Nintedanib effects on tumor cells, besides angiogenesis
Elaine Reguera-Nuñez et al.
Angiogenesis, 22(4), 535-546 (2019-08-30)
In contrast to VEGF pathway-targeting antibodies, antiangiogenic tyrosine kinase inhibitors (TKIs) have failed to meet primary endpoints in almost all phase III clinical trials when combined with conventional chemotherapy. One exception is the combination of nintedanib and docetaxel as a
Frank Hilberg et al.
Cancer research, 68(12), 4774-4782 (2008-06-19)
Inhibition of tumor angiogenesis through blockade of the vascular endothelial growth factor (VEGF) signaling pathway is a novel treatment modality in oncology. Preclinical findings suggest that long-term clinical outcomes may improve with blockade of additional proangiogenic receptor tyrosine kinases: platelet-derived

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service