Skip to Content
Merck
All Photos(1)

Key Documents

SML0603

Sigma-Aldrich

Lixivaptan

≥96% (HPLC)

Synonym(s):

N-[3-Chloro-4-(5H-pyrrolo[2,1-c][1,4]benzodiazepine-10(11H)-ylcarbonyl)phenyl]-5-fluoro-2-methylbenzamide, VPA 985, WAY-VPA 985

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H21ClFN3O2
CAS Number:
Molecular Weight:
473.93
UNSPSC Code:
51111800
NACRES:
NA.77

description

4% w/w in ethyl acetate

Assay

≥96% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

InChI

1S/C27H21ClFN3O2/c1-17-8-9-19(29)13-23(17)26(33)30-20-10-11-22(24(28)14-20)27(34)32-16-21-6-4-12-31(21)15-18-5-2-3-7-25(18)32/h2-14H,15-16H2,1H3,(H,30,33)

InChI key

PPHTXRNHTVLQED-UHFFFAOYSA-N

Biochem/physiol Actions

Arginine vasopressin (AVP) plays an important part in circulatory and water homoeostasis and is important in renal hemodynamic alterations, water retention, and cardiac remodeling in congestive heart failure (CHF). Lixivaptan is an AVP V2-receptor selective antagonist that increases water excretion and normalizes serum sodium levels. Lixivaptan has been in clinical trials for hyponatremia associated with heart failure.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service