Skip to Content
Merck
All Photos(1)

Key Documents

O1016

Sigma-Aldrich

OPC-21268 hydrate

≥98% (HPLC)

Synonym(s):

1-{1-[4(3-acetylaminopropoxy)benzoyl]-4-piperidyl}-3,4-dihydro-2(1H)-quinolinone hydrate, N-[3-[4-[[4-(3,4-dihydro-2-oxo-1(2H)-quinolinyl)-1-piperidinyl]carbonyl]phenoxy]propyl]-acetamide hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H31N3O4 · xH2O
CAS Number:
Molecular Weight:
449.54 (anhydrous basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥20 mg/mL

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

storage temp.

2-8°C

SMILES string

O.CC(=O)NCCCOc1ccc(cc1)C(=O)N2CCC(CC2)N3C(=O)CCc4ccccc34

InChI

1S/C26H31N3O4.H2O/c1-19(30)27-15-4-18-33-23-10-7-21(8-11-23)26(32)28-16-13-22(14-17-28)29-24-6-3-2-5-20(24)9-12-25(29)31;/h2-3,5-8,10-11,22H,4,9,12-18H2,1H3,(H,27,30);1H2

InChI key

QTCWOUIERWBMQV-UHFFFAOYSA-N

Biochem/physiol Actions

OPC-21268 is a vasopressin V1a selective antagonist.
OPC-21268 is one of 2 non-peptide V1a selective antagonists, along with SR-49059. Arginine vasopressin (AVP) is a hormone that plays an important part in circulatory and water homoeostasis and is important in renal hemodynamic alterations, water retention, and cardiac remodeling in congestive heart failure (CHF). There are three AVP receptor subtypes-V1a, V1b, and V2 - all belong to the large rhodopsin-like G-protein-coupled receptor family. V(1a) antagonists improve water balance and cardiac hypertrophy in CHF and might be beneficial for the treatment of water retention and cardiac remodeling in CHF.

Features and Benefits

This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James Walter Ferguson et al.
Clinical science (London, England : 1979), 105(1), 1-8 (2003-03-18)
Vasopressin, or antidiuretic hormone, is a peptide hormone that is released from the posterior pituitary gland in response to changes in blood pressure and plasma osmolality. The main pathophysiological states associated with high plasma vasopressin concentrations are cirrhosis, cardiac failure
Chun-Ping Chu et al.
Neuroscience research, 53(2), 147-155 (2005-07-26)
The present study is an investigation of the responses of the cardiovascular system and Fos expression to intracerebroventricular (i.c.v.) administration of hypertonic saline (HS) in conscious arginine vasopressin (AVP)-overexpressing transgenic (Tg) and control rats. Central HS (0.3, 0.67, or 1.0M
Sho Maejima et al.
General and comparative endocrinology, 157(2), 196-202 (2008-06-17)
Anuran amphibians do not drink orally but absorb water osmotically through the highly permeable ventral skin. In this cutaneous water absorption, roles of the putative cerebral osmoreceptors and functions of arginine vasotocin (AVT) were examined in the central nervous system
Nina Japundzić-Zigon et al.
Journal of pharmacological sciences, 95(1), 47-55 (2004-05-22)
Effects of V(1) (OPC-21268) and V(2) (OPC-31260) vasopressin antagonists on blood pressure (BP) short-term variability were investigated in adult spontaneously hypertensive rats (SHR) under basal conditions and after the stimulation of vasopressin release by hemorrhage. BP was recorded intra-arterially and
Kazushige Tachikawa et al.
American journal of physiology. Endocrinology and metabolism, 285(6), E1161-E1166 (2003-08-14)
Although arginine vasopressin (AVP), an antidiuretic hormone, has been widely acknowledged to play an important role in cardiovascular regulation via V1a receptors (V1aR), its precise significance remains unclear. In this study, we investigated the effects of long-standing high plasma AVP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service