Skip to Content
Merck
All Photos(1)

Documents

SML0322

Sigma-Aldrich

SR1001

≥98% (HPLC)

Synonym(s):

N-(5-(N-(4-(1,1,1,3,3,3-Hexafluoro-2-hydroxypropan-2-yl)phenyl)sulphamoyl)-4-methylthiazol-2-yl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13F6N3O4S2
CAS Number:
Molecular Weight:
477.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >5 mg/mL

storage temp.

2-8°C

SMILES string

CC(=O)Nc1nc(C)c(s1)S(=O)(=O)Nc2ccc(cc2)C(O)(C(F)(F)F)C(F)(F)F

InChI

1S/C15H13F6N3O4S2/c1-7-11(29-12(22-7)23-8(2)25)30(27,28)24-10-5-3-9(4-6-10)13(26,14(16,17)18)15(19,20)21/h3-6,24,26H,1-2H3,(H,22,23,25)

InChI key

OZBSSKGBKHOLGA-UHFFFAOYSA-N

Application

SR1001 has been used as retinoic-acid receptor (RAR)-related orphan nuclear receptor gamma (RORγt) inhibitor to study its effects on interleukin 17 (IL-17) expression in itch cells , as a RAR-related orphan receptor alpha (RORα)- inhibitor to study its effects on RORα-dependent transcription of glucose-6-phosphatase (G6Pase) .
SR1001 has been used for inhibiting retinoid-related orphan receptor-γ t (RORγT) activation in Itch−/− mice.

Biochem/physiol Actions

SR1001 is an antagonist of the nuclear retinoic acid receptor-related orphan receptors RORα and RORγt with no activity at LXR or RORβ. RORα and RORγt are essential for the development of TH17 cells, T-helper cells that produce interleukin-17 and have recently been shown to have pathological roles in various autoimmune diseases. SR1001 binds to the ligand-binding domain of RORα and RORγt to decrease affinity of the receptor for coactivators and increse affinity for co-repressors. It inhibited the differentiation and function of TH17 cells and suppressed the clinical severity of a mouse model of multiple sclerosis.

Features and Benefits

This compound is featured on the Nuclear Receptors (PPARs) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahesh Kathania et al.
Nature immunology, 17(8), 997-1004 (2016-06-21)
Dysregulated expression of interleukin 17 (IL-17) in the colonic mucosa is associated with colonic inflammation and cancer. However, the cell-intrinsic molecular mechanisms by which IL-17 expression is regulated remain unclear. We found that deficiency in the ubiquitin ligase Itch led
The cAMP-dependent protein kinase downregulates glucose-6-phosphatase expression through ROR$\alpha$ and SRC-2 coactivator transcriptional activity
Madsen A, et al.
Molecular and Cellular Endocrinology, 419(8) (2016)
Andre Madsen et al.
Molecular and cellular endocrinology, 419, 92-101 (2015-10-13)
Fasting hormones activate the cAMP/PKA signaling pathway and stimulate expression of hepatic gluconeogenic enzymes including glucose-6-phosphatase (G6Pase). Previously it was shown that steroid receptor coactivator 2 (SRC-2) knock-out mice exhibit fasting hypoglycemia and that SRC-2 coactivates RAR-related orphan receptor alpha
Zhengguang Wang et al.
Oncotarget, 8(7), 11105-11113 (2017-01-05)
Retinoid-related orphan receptor α (RORα) is a nuclear receptor, which regulates inflammation and immune responses, lipid metabolism and circadian rhythm. Although RORα suppresses breast tumor invasion, it is unknown whether RORα is dysregulated in gastric cancer leading to cellular survival.
Patricia R Taylor et al.
Journal of leukocyte biology, 100(1), 213-222 (2016-04-02)
IL-6 and IL-23 (IL-6/23) induce IL-17A (IL-17) production by a subpopulation of murine and human neutrophils, resulting in autocrine IL-17 activation, enhanced production of reactive oxygen species, and increased fungal killing. As IL-6 and IL-23 receptors trigger JAK1, -3/STAT3 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service