Skip to Content
Merck
All Photos(1)

Documents

SML0306

Sigma-Aldrich

Biapenem

≥98% (HPLC)

Synonym(s):

6-[[(4R,5S,6S)-2-Carboxy-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-en-3-yl]thio]-6,7-dihydro-5H-pyrazolo[1,2-a][1,2,4]triazol-4-ium inner salt, Biapenern, CL 186-815, CL 186815, L 627, LJC 10627

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N4O4S
CAS Number:
Molecular Weight:
350.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: ≥5 mg/mL (warmed)

storage temp.

−20°C

SMILES string

C[C@@H](O)[C@@H]1[C@H]2[C@@H](C)C(SC3Cn4cnc[n+]4C3)=C(N2C1=O)C([O-])=O

InChI

1S/C15H18N4O4S/c1-7-11-10(8(2)20)14(21)19(11)12(15(22)23)13(7)24-9-3-17-5-16-6-18(17)4-9/h5-11,20H,3-4H2,1-2H3/t7-,8-,10-,11-/m1/s1

InChI key

MRMBZHPJVKCOMA-YJFSRANCSA-N

Biochem/physiol Actions

Biapenem does not react to renal dihydropeptidase-I mediated hydrolysis in human. Biapenem is well absorbed by tissues and body fluids, following intravenous injection.
Biapenem is a broad spectrum, carbapenem-based antibiotic with activity against both Gram-positive and Gram-negative bacterial strains.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabio Simona et al.
The Journal of biological chemistry, 284(41), 28164-28171 (2009-08-13)
Metallo-beta-lactamases (MbetaLs) constitute an increasingly serious clinical threat by giving rise to beta-lactam antibiotic resistance. They accommodate in their catalytic pocket one or two zinc ions, which are responsible for the hydrolysis of beta-lactams. Recent x-ray studies on a member
Eiki Kikuchi et al.
Antimicrobial agents and chemotherapy, 53(7), 2799-2803 (2009-04-22)
The time above the MIC (T>MIC) is the pharmacokinetic/pharmacodynamic (PK/PD) parameter that correlates with the therapeutic efficacy of beta-lactam antibiotics. A prolonged infusion can provide plasma drug concentrations that remain above the MIC for a long period. The objective of
Antonella R Palacios et al.
Biomolecules, 10(6) (2020-06-07)
β-Lactam antibiotics are the most widely prescribed antibacterial drugs due to their low toxicity and broad spectrum. Their action is counteracted by different resistance mechanisms developed by bacteria. Among them, the most common strategy is the expression of β-lactamases, enzymes
Darija Viducic et al.
Antimicrobial agents and chemotherapy, 60(10), 5752-5764 (2016-07-20)
The ability of Pseudomonas aeruginosa to rapidly modulate its response to antibiotic stress and persist in the presence of antibiotics is closely associated with the process of cell-to-cell signaling. The alternative sigma factor RpoN (σ(54)) is involved in the regulation
Sharon H Ackerman et al.
PloS one, 8(1), e55136-e55136 (2013-02-02)
A general mechanism has been proposed for metallo β-lactamases (MβLs), in which deprotonation of a water molecule near the Zn ion(s) results in the formation of a hydroxide ion that attacks the carbonyl oxygen of the β-lactam ring. However, because

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service