Skip to Content
Merck
All Photos(1)

Key Documents

SML0031

Sigma-Aldrich

DBeQ

≥98% (HPLC)

Synonym(s):

JRF 12, N2,N4-dibenzylquinazoline-2,4-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20N4
CAS Number:
Molecular Weight:
340.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥20 mg/mL

storage temp.

room temp

SMILES string

C(Nc1nc(NCc2ccccc2)c3ccccc3n1)c4ccccc4

InChI

1S/C22H20N4/c1-3-9-17(10-4-1)15-23-21-19-13-7-8-14-20(19)25-22(26-21)24-16-18-11-5-2-6-12-18/h1-14H,15-16H2,(H2,23,24,25,26)

InChI key

QAIMUUJJAJBPCL-UHFFFAOYSA-N

Application

HeLa cells were treated with DBeQ and the effects on in vivo ubiquitination and protein dislocation were studied by live cell imaging.

Biochem/physiol Actions

ATPase p97 inhibitor.
DBeQ is a potent and specific inhibitor of ATPase p97, an integral component of the ubiquitin-fusion degradation (UFD) pathway. DBeQ inhibits the degradation of ubiquitinated proteins, the endoplasmic reticulum-associated degradation pathway, and autophagosome maturation. The compound also potently inhibits cellular proliferation and induces caspase 3/7 activity and apoptosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ainoa Figuerola-Conchas et al.
ACS chemical biology, 15(1), 243-253 (2019-12-04)
VCP/p97 belongs to the AAA+ ATPase family and has an essential role in several cellular processes ranging from cell division to protein homeostasis. Compounds targeting p97 inhibit the main ATPase domain and cause cell death. Here, using PNA-encoded chemical libraries
Holger W Auner et al.
PloS one, 8(9), e74415-e74415 (2013-09-27)
Inhibition of the proteasome is a widely used strategy for treating multiple myeloma that takes advantage of the heavy secretory load that multiple myeloma cells (MMCs) have to deal with. Resistance of MMCs to proteasome inhibition has been linked to
Wenchun Fan et al.
Antiviral research, 127, 32-40 (2016-01-19)
The tripartite motif protein 21 (TRIM21) is a ubiquitously expressed E3 ubiquitin ligase and an intracellular antibody receptor. TRIM21 mediates antibody-dependent intracellular neutralization (ADIN) in cytosol and provides an intracellular immune response to protect host defense against pathogen infection. In
Aparna Shinde et al.
Cancer research, 79(8), 1831-1843 (2019-02-09)
The ability of breast cancer cells to transiently transition between epithelial and mesenchymal states contributes to their metastatic potential. Therefore, driving tumor cells into a stable mesenchymal state, as opposed to complete tumor cell eradication, presents an opportunity to pharmacologically
Wei-Wei Luo et al.
PLoS pathogens, 13(11), e1006693-e1006693 (2017-11-21)
VISA (also known as MAVS, IPS-1 and Cardif) is an essential adaptor protein in innate immune response to RNA virus. The protein level of VISA is delicately regulated before and after viral infection to ensure the optimal activation and timely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service