Skip to Content
Merck
All Photos(1)

Key Documents

SML1071

Sigma-Aldrich

ML240

≥98% (HPLC)

Synonym(s):

2-(2-Amino-1H-benzimidazol-1-yl)-8-methoxy-N-(phenylmethyl)-4-quinazolinamine, CID-49830258

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H20N6O
CAS Number:
Molecular Weight:
396.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

, white to yellow to light brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

NC1=NC2=C(C=CC=C2)N1C3=NC4=C(OC)C=CC=C4C(NCC5=CC=CC=C5)=N3

InChI

1S/C23H20N6O/c1-30-19-13-7-10-16-20(19)27-23(28-21(16)25-14-15-8-3-2-4-9-15)29-18-12-6-5-11-17(18)26-22(29)24/h2-13H,14H2,1H3,(H2,24,26)(H,25,27,28)

InChI key

NHAMBLRUUJAFOY-UHFFFAOYSA-N

Application

ML240 has been used as an inhibitor of D2 ATPase activity of valosin-containing protein.

Biochem/physiol Actions

ML240 is a selective, ATP-competetive inhibitor of AAA ATPase p97, also called valosine containing protein (VCP).
ML240 is a selective, ATP-competetive inhibitor of AAA ATPase p97, also called valosine containing protein (VCP). The IC50 for p97 inhibition is 110 nM. ML240 rapidly induces activity of Caspases 3 and 7, leading to apoptosis. ML240 has potent antiproliferative activity towards NCI-60 cancer cell lines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Specific mutations in the D1?D2 linker region of VCP/p97 enhance ATPase activity and confer resistance to VCP inhibitors.
Bastola P, et al.
Cell Death & Disease, 3, 17065-17065 (2017)
Feng Wang et al.
ChemMedChem, 15(8), 685-694 (2020-03-13)
A major challenge of targeted cancer therapy is the selection for drug-resistant mutations in tumor cells leading to loss of treatment effectiveness. p97/VCP is central regulator of protein homeostasis and a promising anticancer target because of its vital role in
Pratikkumar H Vekaria et al.
Leukemia, 33(7), 1675-1686 (2019-01-22)
p97 is an ATPase that works in concert with histone deacetylase 6 (HDAC6), to facilitate the degradation of misfolded proteins by autophagosomes. p97 has also been implicated in DNA repair and maintaining genomic stability. In this study, we determined the
Jasmine Harley et al.
Brain communications, 3(3), fcab166-fcab166 (2021-08-17)
RNA binding proteins have been shown to play a key role in the pathogenesis of amyotrophic lateral sclerosis (ALS). Mutations in valosin-containing protein (VCP/p97) cause ALS and exhibit the hallmark nuclear-to-cytoplasmic mislocalization of RNA binding proteins (RBPs). However, the mechanism
Richard S Marshall et al.
Cell, 177(3), 766-781 (2019-04-09)
During autophagy, vesicle dynamics and cargo recruitment are driven by numerous adaptors and receptors that become tethered to the phagophore through interactions with lipidated ATG8/LC3 decorating the expanding membrane. Most currently described ATG8-binding proteins exploit a well-defined ATG8-interacting motif (AIM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service