Skip to Content
Merck
All Photos(1)

Key Documents

SMB00810

Sigma-Aldrich

Oleocanthal

from Olea europaea

Synonym(s):

(-)-Oleocanthal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20O5
CAS Number:
Molecular Weight:
304.34
UNSPSC Code:
12352205
NACRES:
NA.28

biological source

Olea europaea

Assay

≥95%

form

film

mol wt

304.34

solubility

DMSO: 1 mg/mL, clear, colorless

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

shipped in

dry ice

storage temp.

−20°C

SMILES string

O(CCc1ccc(cc1)O)C(=O)C[C@H](CC=O)\C(=C/C)\C=O

InChI

1S/C17H20O5/c1-2-14(12-19)15(7-9-18)11-17(21)22-10-8-13-3-5-16(20)6-4-13/h2-6,9,12,15,20H,7-8,10-11H2,1H3/b14-2-/t15-/m0/s1

InChI key

VPOVFCBNUOUZGG-VAKDEWRISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oleocanthal, a secoiridoid, a cyclopentane monoterpene, and a member of phenols, is a bioactive natural compound commonly derived from Olive oil (Olea europaea). Current research suggests that this metabolite acts as an inhibitor and may exhibit diverse biological activities, including anti-inflammatory, neuroprotective, apoptosis-inducing, antioxidant, antimalarial, and antineoplastic properties.

Application

Oleocanthal is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and drug discovery research.

Biochem/physiol Actions

Oleocanthal is one of the important polyphenols of olive oil well-known for its biological properties and health benefits. It exerts anti-inflammatory activity due to its dose-dependent inhibition of cyclooxygenase (COX-1, COX-2) enzymes in the prostaglandin biosynthesis in the same pathway as Ibuprofen. Oleocanthal is a potent anti-cancer agent, its treatment demonstrates inhibition of proliferation, migration, and invasion of various human breast cancer, prostate cancer, and multiple myeloma cells. In non-melanoma skin cancer, it reduces cell viability and migration, through inhibition of Erk and Akt phosphorylation and suppression of B-Raf expression. Oleocanthal was found to modulate estrogen receptor expression in luminal breast cancer and acts synergistically with tamoxifen treatment. It also elicits neuroprotective properties by inhibiting the formation of neurofibrillary tangles, a key hallmark in the pathogenesis of Alzheimer′s disease. Oleocanthal has also been reported to increase Amyloid-β transport across the blood-brain barrier via P-glycoprotein induction. It was found that it acts synergistically with Donepezil, an acetylcholine esterase inhibitor approved for use for all Alzheimer′s disease stages. Oleuropein displays anti-leishmanial activity when added in vitro and in vivo, in experimental models of Leishmania infection.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alaa H Abuznait et al.
ACS chemical neuroscience, 4(6), 973-982 (2013-02-19)
Oleocanthal, a phenolic component of extra-virgin olive oil, has been recently linked to reduced risk of Alzheimer's disease (AD), a neurodegenerative disease that is characterized by accumulation of β-amyloid (Aβ) and tau proteins in the brain. However, the mechanism by
Limor Goren et al.
PloS one, 14(8), e0216024-e0216024 (2019-08-15)
(-)-Oleocanthal (oleocanthal) is a phenolic compound found in varying concentrations in extra virgin olive oil oleocanthal has been shown to be active physiologically, benefiting several diseased states by conferring anti-inflammatory and neuroprotective benefits. Recently, we and other groups have demonstrated
Kok-Lun Pang et al.
Nutrients, 10(5) (2018-05-08)
Oleocanthal is a minor constituent of olive oil with strong anti-inflammatory activities. Since the pathogenesis of many chronic diseases involves inflammatory and oxidative components, oleocanthal is a promising agent to prevent these conditions. This review aimed to summarise the current
Gary K Beauchamp et al.
Nature, 437(7055), 45-46 (2005-09-02)
Newly pressed extra-virgin olive oil contains oleocanthal--a compound whose pungency induces a strong stinging sensation in the throat, not unlike that caused by solutions of the non-steroidal anti-inflammatory drug ibuprofen. We show here that this similar perception seems to be
Mohamed R Akl et al.
PloS one, 9(5), e97622-e97622 (2014-05-23)
Dysregulation of the Hepatocyte growth factor (HGF)/c-Met signaling axis upregulates diverse tumor cell functions, including cell proliferation, survival, scattering and motility, epithelial-to-mesenchymal transition (EMT), angiogenesis, invasion, and metastasis. (-)-Oleocanthal is a naturally occurring secoiridoid from extra-virgin olive oil, which showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service