Skip to Content
Merck
All Photos(2)

Documents

SL2

Sigma-Aldrich

Sigmacote®

siliconizing reagent for glass and other surfaces

Synonym(s):

chlorinated organopolysiloxane in heptane, siliconizing reagent for glass

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
NACRES:
NA.47

biological source

synthetic

color

colorless

density

0.68 g/cm3

application(s)

hematology
histology

storage temp.

2-8°C

General description

Sigmacote® is a solution of a chlorinated organopolysiloxane in heptane that readily forms a covalent, microscopically thin film on glass. The film repels water, retards the clotting of blood or plasma, and prevents surface adsorption of many basic proteins. Ready to use without dilution; reusable if kept free of moisture.

Application

Sigmacote® has been used to prevent adhesion of the growth factors to plasticware. It has also been used to coat coverslip in order to prevent the adhesion of larval brain during squashing.

Biochem/physiol Actions

Sigmacote® is ideal for glass, ceramics and fiber optics. It is used to treat GC injection glass inserts.

Legal Information

Sigmacote is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative analysis of satellite DNA in the Drosophila melanogaster species complex
Jagannathan M, et al.
G3 (Bethesda, Md.), g3-116 (2016)
Analysis of protein S-acylation by gas chromatography-coupled mass spectrometry using purified proteins
Sorek, Nadav and Yalovsky, Shaul
Nature Protocols, 5(5), 834-834 (2010)
Isolation of adult stem cells and their differentiation to Schwann cells
Progenitor Cells, 47-57 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service