Skip to Content
Merck
All Photos(2)

Key Documents

6028

Millipore

Amicon® Stirred Cell Reservoir

Synonym(s):

Amicon Cell Reservoir, Cell Reservoir, Stirred Cell Reservoir

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116500
eCl@ss:
32019190

material

Nylon fittings (for tube)
acrylic body
glass-filled nylon stand (retaining)
nitrile O-ring (for relieve valve)
nitrile fittings (for tube)
polyacetal base
polyacetal cap
polyethylene tubing
silicone O-ring (for reservoir)
stainless steel fittings (for tube)

Quality Level

technique(s)

buffer exchange: suitable

shipped in

ambient

General description

The Amicon® Stirred Cell Reservoir automatically replenishes liquid in the stirred cell due to the pressure differential between two vessels, created by an external gas source.

Application

Amicon® Stirred Cell Reservoir has been used in desalination simulator to adjust volume re-striction and enable sample feeding to the cell. It might also be used in stirred membrane filtration.

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modeling process dynamics using a novel neural network architecture: Application to stirred cell microfiltration
Mhurchu JN, et al.
Chemical Engineering Communications , 197, 1152-1162 (2010)
The influence of ion identity and ionic strength on membrane biofouling of a binary protein solution
Kilmer NT, et al.
Separation and Purification Technology, 255, 117769-117769 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service