Skip to Content
Merck
All Photos(1)

Documents

SHC003V

Sigma-Aldrich

MISSION® pLKO.1-puro-CMV-TurboGFP Positive Control Transduction Particles

Green fluorescent protein marker to monitor transduction efficiency

Synonym(s):

MISSION®, MISSION® TurboGFP Control Transduction Particles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

Quality Level

product line

MISSION®

concentration

≥1x106 VP/ml (via p24 assay)

technique(s)

capture ELISA: 106 TU/mL using p24

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

This construct aids in interpretation of experimental design and results by providing a green fluorescent protein marker for monitoring success in transduction of your cells of interest.

TurboGFP is an improved variant of the green fluorescent protein copGFP cloned from the copepoda Pontellina plumata. The TurboGFP transduction particles are produced from the sequence-verified lentiviral plasmid, pLKO.1-puro-CMV-TurboGFP (SHC003). It is a positive control to monitor transduction efficiency.

Self-inactivating replication incompetent viral particles are produced in packaging cells (HEK293T) by co-transfection with compatible packaging plasmids. In addition, the Control Transduction Particles are pseudotyped with an envelope G glycoprotein from Vesicular Stomatitis Virus (VSV-G), allowing transduction of a wide variety of mammalian cells. 200 μl of 106 TU/ml (via p24 titering assay) lentiviral particles are provided as frozen stock.
When conducting experiments using MISSION® shRNA clones, the proper controls should be a key element of your experimental design to allow for accurate interpretation of knockdown results. The MISSION Control Transduction Particles are a critical positive control to monitor transduction efficiency.
To see more application data, protocols, vector maps visit sigma.com/shrna.

Application

MISSION® pLKO.1-puro-CMV-TurboGFP Positive Control Transduction Particles has been used to transduce HCT116 (human colon carcinoma) cell line for 2D culture and to form 3D spheroids. It has also been used to generate fluorescent cell lines by transduction.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
TurboBeads is a trademark of TurboBeads LLC
TurboGFP is a trademark of Evrogen Co.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Two-Photon Microscopy Analysis of Gold Nanoparticle Uptake in 3D Cell Spheroids.
Rane TD and Armani AM
PLoS ONE, 11(12) (2016)
Ruoxiang Wang et al.
The Prostate, 80(3), 274-283 (2019-12-18)
We previously determined that cancer-stromal interaction was a direct route to tumor cell heterogeneity progression, since cancer-stromal cell fusion in coculture resulted in the creation of heterogeneous clones of fusion hybrid progeny. In this report, we modified the cancer-stromal coculture
Esperanza Martín-Sánchez et al.
PloS one, 9(11), e112148-e112148 (2014-11-12)
Currently, there is no efficient therapy for patients with peripheral T cell lymphoma (PTCL). The Proviral Integration site of Moloney murine leukemia virus (PIM) kinases are important mediators of cell survival. We aimed to determine the therapeutic value of PIM
Chi-Li Chiu et al.
Scientific reports, 6, 22435-22435 (2016-03-05)
The androgen receptor (AR) pathway plays a central role in prostate cancer (PCa) growth and progression and is a validated therapeutic target. In response to ligand binding AR translocates to the nucleus, though the molecular mechanism is not well understood.
A high-content image-based method for quantitatively studying context-dependent cell population dynamics.
Garvey CM
Scientific Reports, 6, 29752-29752 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service