Skip to Content
Merck
All Photos(1)

Key Documents

SHC012V

Sigma-Aldrich

MISSION® pLKO.1-puro-CMV-TagRFP Positive Control Transduction Particles

Contains a gene encoding TagRFP

Synonym(s):

MISSION®, MISSION® Control Transduction Particles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

Quality Level

product line

MISSION®

concentration

≥1x106 VP/ml (via p24 assay)

technique(s)

capture ELISA: 106 TU/mL using p24

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

Self-inactivating replication incompetent viral particles are produced in packaging cells (HEK293T) by co-transfection with compatible packaging plasmids. In addition, the Control Transduction Particles are pseudotyped with an envelope G glycoprotein from Vesicular Stomatitis Virus (VSV-G), allowing transduction of a wide variety of mammalian cells. 200 μl of 106 TU/ml (via p24 titering assay) lentiviral particles are provided as frozen stock.
When conducting experiments using MISSION® shRNA clones, the proper controls should be a key element of your experimental design to allow for accurate interpretation of knockdown results. The MISSION Control Transduction Particles are a critical positive control to monitor transduction efficiency.
To see more application data, protocols, vector maps visit sigma.com/shrna.

Application

MISSION® pLKO.1-puro-CMV-TagRFP Positive Control Transduction Particles has been used to knockdown transient receptor potential vanilloid 1 receptor (TRPV1)-like receptors. It has also been used to generate fluorescent cell lines.
MISSION®pLKO.1-puro-CMV-TagRFP Positive Control Transduction Particles has been used in generating fluorescent cell lines by transduction. It has also been used in transfection of human dermal fibroblast (HDF), WI-38 and IMR-90 FBs to express tagRFP.
To see more application data, protocols, vector maps visit sigma.com/shrna.
When conducting experiments using MISSION shRNA clones, the proper controls should be a key element of your experimental design to allow for accurate interpretation of knockdown results. The MISSION TagRFP Control Transduction Particles are a positive control to monitor transduction efficiency. The MISSION TagRFP Control Transduction Particles are produced from the sequence-verified 8594 base pair lentivirus plasmid, pLKO.1-puro-CMV-TagRFP (SHC012).

This construct aids in interpretation of experimental design and results by providing a red fluorescent protein marker for monitoring success in transduction of your cells of interest.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
TagRFP is a trademark of Evrogen Co.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High EMT signature score of invasive non-small cell lung cancer (NSCLC) cells correlates with NF kappa B driven colony-stimulating factor 2 (CSF2/GM-CSF) secretion by neighboring stromal fibroblasts
Rudisch A, et al.
PLoS ONE, 10(4), e0124283-e0124283 (2015)
Activation of temperature-sensitive TRPV1-like receptors in ARC POMC neurons reduces food intake
Jeong JH, et al.
PLoS Biology, 16(4), e2004399-e2004399 (2018)
A high-content image-based method for quantitatively studying context-dependent cell population dynamics.
Garvey CM
Scientific Reports, 6, 29752-29752 (2016)
Albin Rudisch et al.
PloS one, 10(4), e0124283-e0124283 (2015-04-29)
We established co-cultures of invasive or non-invasive NSCLC cell lines and various types of fibroblasts (FBs) to more precisely characterize the molecular mechanism of tumor-stroma crosstalk in lung cancer. The HGF-MET-ERK1/2-CREB-axis was shown to contribute to the onset of the
Jae Hoon Jeong et al.
PLoS biology, 16(4), e2004399-e2004399 (2018-04-25)
Proopiomelanocortin (POMC) neurons in the arcuate nucleus of the hypothalamus (ARC) respond to numerous hormonal and neural signals, resulting in changes in food intake. Here, we demonstrate that ARC POMC neurons express capsaicin-sensitive transient receptor potential vanilloid 1 receptor (TRPV1)-like

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service