Skip to Content
Merck
All Photos(1)

Documents

SHCLNV

Sigma-Aldrich

MISSION® shRNA

Lentiviral Particles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106609

technique(s)

capture ELISA: 106 VP/mL using p24

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

Our comprehensive shRNA product offering consists of over 150,000 pre-cloned shRNA constructs targeting more than 15,000 human and 15,000 mouse genes. The library is available in: gene family sets: gene collections related to specific functional classes such as kinases and ion channels; gene taget sets, sets of shRNA clones that target your favorite gene; and individual shRNA clones. Our shRNA and lentiviral manufacturing platforms utilize dedicated laboratories featuring state-of-the-art robotics, liquid handling and laboratory information management systems (LIMS) to provide superior quality and service for your RNAi research needs.
Lentiviral particles are ideal for transducing a wide range of cell lines. VSV-G Psuedotyping of the MISSION® lentiviral particles allows for entry into a wide range of cells. In addition, lentiviral particles stably integrate the shRNA into the genomes of both dividing and non-dividing cell lines. The MISSION bioproduction team regularly produces high quality lentivirus, so that our customers can get straight to their RNAi experiment without wasting precious time on tedious lentiviral production. Our minimum titer is 106 VP/mL. Unlike adenoviral infections that require vast excesses, lentiviral transduction is efficient enought allow for MOI as low as 1.

Application

MISSION® shRNA has been used:
  • in transfections and transductions
  • in the establishment of stable cell lines expressing shRNA
  • in the generation of alpha-actinin-4 (ACTN4) knockdown cell line
  • to transduce raw 264.7 cells

Other Notes

To see protocols and application data please visit sigma.com/shrna.

Legal Information

Use of this product is subject to one or more license agreements.
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

alpha-Actinin-4 confers radioresistance coupled invasiveness in breast cancer cells through AKT pathway
Desai S, et al.
Biochimica et Biophysica Acta - Molecular Cell Research, 1865(1), 196-208 (2018)
Bile acids reduce endocytosis of high-density lipoprotein (HDL) in HepG2 cells.
Rohrl C, Eigner K, Fruhwurth S, et al.
PLoS ONE, 9(7), e102026-e102026 (2014)
Barbara D Boyan et al.
Scientific reports, 8(1), 8588-8588 (2018-06-07)
Successful osseointegration of an endosseous implant involves migration and differentiation of mesenchymal stem cells (MSCs) on the implant surface. Micro-structured, hydrophilic titanium surfaces direct MSCs to undergo osteoblastic differentiation in vitro, in the absence of media additives commonly used in
Vera Mugoni et al.
Cell research, 29(6), 446-459 (2019-04-27)
Although targeted therapies have proven effective and even curative in human leukaemia, resistance often ensues. IDH enzymes are mutated in ~20% of human AML, with targeted therapies under clinical evaluation. We here characterize leukaemia evolution from mutant IDH2 (mIDH2)-dependence to
Sejal Desai et al.
Biochimica et biophysica acta, 1865(1), 196-208 (2017-10-23)
Acquired radioresistance accompanied with increased metastatic potential is a major hurdle in effective radiotherapy of breast cancers. However, the nature of their inter-dependence and the underlying mechanism remains largely intangible. By employing radioresistant (RR) cell lines, we herein demonstrate that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service