Skip to Content
Merck
All Photos(4)

Key Documents

SAB4200701

Sigma-Aldrich

Anti-CRISPR/Cas9 antibody, Mouse monoclonal

clone 7A9-3A3, purified from hybridoma cell culture

Synonym(s):

CRISPR-associated protein-9 nuclease, Crisper, Crispr RNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

7A9-3A3, monoclonal

form

buffered aqueous solution

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

immunoblotting: 1-2 μg/mL using whole extracts of human HEK-293T cells over-expressing CAS9 protein
immunofluorescence: 1-2 μg/mL using human HEK-293T cells over-expressing CAS9 protein.
immunoprecipitation (IP): 5-10 μg using whole extract of human HEK-293T cells over-expressing CAS9 protein

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

CAS9, also known as clustered regularly interspaced short palindromic repeats (CRISPR)-associated protein-9 nuclease , cas5 and csn1, is the signature gene of the type II CRISPR -RuvC (RNase H-like fold) cas system. CAS9 contains 1388 amino acids. This protein is predicted to contain a RuvC/ ribonuclease (RNase) H domain involved in crRNA maturation and McrA/HNH signature domain involved in the DNA degradation step.
CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats) belongs to the type II CRISPR/CAS9 system. It is part of an adaptive immune system of the Streptococcus pyogenes SF370, protecting from pathogens targets genes by cleaving the foreign DNA in a sequence-dependent manner.2 The type II CRISPR/Cas system which has been adapted to expression in eukaryotic cells, consists of four genes including the Cas9 (CRISPR-associated proteins) nuclease, two noncoding CRISPR RNAs (crRNAs, or gRNA), trans-activating crRNA (tracrRNA) and a precursor crRNA (pre-crRNA) array. The pre-crRNA contains nuclease guide sequences (spacers) interspaced by identical direct repeats (DRs).
The Cas9 endonuclease can be engineered with a single gRNA, directing a DNA double-strand break (DSB) at a desired genomic location. Similar to DSBs induced by zinc finger nucleases (ZFNs), the cell then activates endogenous DNA repair processes, either non-homologous end joining (NHEJ) or homology-directed repair (HDR), to heal the targeted DSB. In comparison to other genome-editing technologies such as designer zinc fingers (ZFs), transcription activator–like effectors (TALEs) and homing meganucleases, the CRISPR/CAS9 system is a scalable, affordable and easy to engineer. Therefore, the anti-CRISPR/CAS9 antibody can be a useful tool for detecting CRISPR/CAS9 positively transfected cells, reveling DSB sites in the genome and in ChIP (Chromatin Immunoprecipitation) related assays.

Immunogen

a recombinant protein within the N-terminal region of Streptococcus pyogene Cas9

Application

Monoclonal Anti-CRISPR/CAS9 recognizes over-expression systems containing CAS9 construct. The antibody successfully recognizes mutant Cas9 variants, including nickase Cas9 and dead Cas9 (dCas9). The antibody may be used in various immunochemical techniques including Immunoblotting, Immunofluorescence and Immunoprecipitation. Monoclonal Anti-CRISPR/CAS9 does not cross react with FnCas9 from Francisella novicida bacteria and Cpf1 proteins from Acidaminococcus sp. (strain BV3L6) and Lachnospiraceae bacterium ND2006.

Biochem/physiol Actions

CAS9 plays a vital role in plasmid DNA interference. It is the only cas protein needed to deliver resistance against foreign DNA. CAS9 stimulates both RNA-guided genome editing and gene regulation in various organisms, but it can facilitate only one activity at a time within any given cell.

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Other Notes

This product is for R&D use only, not for drug, household, or other uses.
In order to obtain best results in different techniques and preparations we recommend determining optimal working concentration by titration test.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L G V Fernandes et al.
Scientific reports, 9(1), 1839-1839 (2019-02-14)
Leptospirosis is a worldwide zoonosis caused by pathogenic bacteria of the genus Leptospira, which also includes free-living saprophyte strains. Many aspects of leptospiral basic biology and virulence mechanisms remain unexplored mainly due to the lack of effective genetic tools available
Jana Ordon et al.
Plant methods, 19(1), 30-30 (2023-03-29)
In plant genome editing, RNA-guided nucleases such as Cas9 from Streptococcus pyogenes (SpCas9) predominantly induce small insertions or deletions at target sites. This can be used for inactivation of protein-coding genes by frame shift mutations. However, in some cases, it
Alicia Davis et al.
Molecular therapy. Methods & clinical development, 25, 158-169 (2022-04-12)
Hypoxia is a characteristic feature of solid tumors that contributes to tumor aggressiveness and is associated with resistance to cancer therapy. The hypoxia inducible factor-1 (HIF-1) transcription factor complex mediates hypoxia-specific gene expression by binding to hypoxia-responsive element (HRE) sequences
Yunjian Wu et al.
Molecular oncology, 17(3), 469-486 (2023-01-07)
Reciprocal interactions between prostate cancer cells and carcinoma-associated fibroblasts (CAFs) mediate cancer development and progression; however, our understanding of the signalling pathways mediating these cellular interactions remains incomplete. To address this, we defined secretome changes upon co-culture of prostate epithelial
Yen-Ting Liu et al.
Molecular and cellular biology, 43(3), 115-129 (2023-03-22)
CDKN2A/B deletion or silencing is common across human cancer, reinforcing the general importance of bypassing its tumor suppression in cancer formation or progression. In rhabdomyosarcoma (RMS) and neuroblastoma, two common childhood cancers, the three CDKN2A/B transcripts are independently expressed to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service