Skip to Content
Merck
All Photos(2)

Documents

SAB4502861

Sigma-Aldrich

Anti-Somatostatin antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Growth hormone release-inhibiting factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 12 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SST(6750)

General description

Anti-Somatostatin antibody detects endogenous levels of total Somatostatin protein.
The SST (somatostatin) gene codes for a peptide hormone containing 14 and 28 amino acid moieties. SST-14 is highly expressed in the central nervous system and peripheral organs. SST-28 is mainly expressed in enteroendocrine cells of the intestine.

Immunogen

The antiserum was produced against synthesized peptide derived from human Somatostatin.

Immunogen Range: 10-59

Application

Anti-Somatostatin, N-Terminal antibody produced in rabbit has been used in immunofluorescence.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Somatostatin plays an important role in regulating the metabolism of other hormones such as growth hormone, insulin, IGF-1 (insulin-like growth factor 1), glucagon, glucagon-like peptide and gastrin. Somatostatin has both regulatory and inhibitory function. It retards gastrointestinal motility and gallbladder contraction, suppresses gut exocrine secretion and reduces epithelial proliferation. Somatostatin is associated with intestinal nutrient absorption and blood flow regulation. This peptide hormone is believed to inhibit cell proliferation and promote apoptosis. Somatostatin acts as a neurotransmitter and neuromodulator. It is associated in maintaining water balance.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Immunofluorescent visualization of mouse interneuron subtypes.
Molgaard S
F1000Research, 3:242, 1-17 (2014)
Use of the Fluidigm C1 platform for RNA sequencing of single mouse pancreatic islet cells.
Xin Y
Proceedings of the National Academy of Sciences of the USA, 113(12), 3293-3298 (2016)
Rulan Yi et al.
CNS neuroscience & therapeutics, 30(6), e14782-e14782 (2024-06-03)
The thalamus system plays critical roles in the regulation of reversible unconsciousness induced by general anesthetics, especially the arousal stage of general anesthesia (GA). But the function of thalamus in GA-induced loss of consciousness (LOC) is little known. The thalamic
Promoter hypermethylation-related reduced somatostatin production promotes uncontrolled cell proliferation in colorectal cancer.
Leiszter K
PLoS ONE, 10(2), 1-17 (2015)
N-Terminal Prosomatostatin as a Risk Marker for Cardiovascular Disease and Diabetes in a General Population.
Hedback T
The Journal of Clinical Endocrinology and Metabolism, 101(9), 3437-3444 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service