Skip to Content
Merck
All Photos(1)

Key Documents

I7633

Sigma-Aldrich

IMDM

With ʟ-glutamine and HEPES, without sodium bicarbonate, powder, suitable for cell culture

Synonym(s):

Iscove′s MDM, Iscoves MDM, Iscove’s Modified Dulbecco’s Medium, IMDM

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Product Name

Iscove′s Modified Dulbecco′s Medium, With L-glutamine and 25 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture

form

powder

technique(s)

cell culture | hybridoma: suitable
cell culture | mammalian: suitable

components

phenol red: yes
glucose: yes
L-glutamine: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Iscove′s Modified Dulbecco′s medium is a modification of Dulbecco′s Modified Eagle′s Medium (DME) containing selenium, additional amino acids, and vitamins, sodium pyruvate, 4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid (HEPES) buffer, and potassium nitrate instead of ferric nitrate. Iscove′s medium supports murine B lymphocytes, hemopoietic tissue from bone marrow, B cells stimulated with lipopolysaccharide, T lymphocytes, and a variety of hybrid cells.

Application

Iscove′s Modified Dulbecco′s Medium has been used to culture KG1a cells (human leukemic cell line) and Chinese hamster ovary (CHO) DG44 cell line.
Reported to support murine B lymphocytes, hematopoietic tissue from bone marrow, B cells stimulated with lipopolysaccharide, T lymphocytes, and a variety of hybrid cells.

Quantity

Formulated to contain 17.7 grams of powder per liter of medium.

Reconstitution

Supplement with 3.024 g/L sodium bicarbonate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth A Henderson et al.
Virology journal, 17(1), 43-43 (2020-04-03)
Zika virus (ZIKV) is a major human pathogen and member of the Flavivirus genus. Previous studies have identified neutralizing antibodies from Zika patients that bind to quaternary epitopes across neighboring envelope (E) proteins, called E dimer epitopes (EDE). An asparagine-linked
T Yoshikawa et al.
Cytotechnology, 33(1-3), 37-46 (2008-11-13)
In order to establish an easy and quick construction method for obtaining a stable and highly productive gene-amplified recombinant Chinese Hamster Ovary (CHO) cell line, variouskinds of stepwise methotrexate (MTX) selection were carriedout. The specific growth and production rates of
Hale M Tasyurek et al.
Human gene therapy, 29(7), 802-815 (2018-02-08)
Postprandial glucose-induced insulin secretion from the islets of Langerhans is facilitated by glucagon-like peptide-1 (GLP-1)-a metabolic hormone with insulinotropic properties. Among the variety of effects it mediates, GLP-1 induces delta cell secretion of somatostatin, inhibits alpha cell release of glucagon
Ryan S Pawell et al.
Biomicrofluidics, 7(5), 56501-56501 (2014-01-10)
Deterministic lateral displacement (DLD) is a microfluidic size-based particle separation or filter technology with applications in cell separation and enrichment. Currently, there are no cost-effective manufacturing methods for this promising microfluidic technology. In this fabrication paper, however, we develop a
Yogesh Kumar Verma et al.
International journal of radiation biology, 96(5), 671-688 (2020-01-28)
Purpose: Lithium chloride (LiCl) is clinically used for manic disorders. Its role has been shown in improving cell survival by decreasing Bax and p53 expression and increasing Bcl-2 concentration in the cell. This potential of LiCl is responsible for reducing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service