Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200039

Sigma-Aldrich

Anti-Serotonin Transporter (N-terminal) antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-5-hydroxytryptamine transporter, Anti-5HTT, Anti-HTT, Anti-OCD1, Anti-SERT, Anti-SLC6A4 solute carrier family 6, member 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~83 kDa

species reactivity

human

concentration

~1.5 mg/mL

technique(s)

western blot: 1.5-3.0 μg/mL using SH-SY5Y cell lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Solute carrier family 6 member 4 (SLC6A4), also known as 5-hydroxytryptamine transporter (5-HTT), is an integral membrane protein, encoded by the gene mapped to human chromosome 17q11.2. The encoded protein is a member of SLC6 gene family and is mainly expressed in brain and blood cells.

Application

Anti-Serotonin Transporter (N-terminal) antibody produced in rabbit has been used in western blot analysis and immunohistochemistry.

Biochem/physiol Actions

Solute carrier family 6 member 4 (SLC6A4) catalyzes the transport of neurotransmitter serotonin from the synaptic space into presynaptic neurons. It also plays a vital role in mood and anxiety regulation. Mutation on the gene might alter the expression of serotonin (5HT) in the synaptic cleft, which ultimately leads to schizophrenia. Allelic variations in the gene increases the risk of susceptibility to autism and rigid-compulsive behaviors.
The polymorphic promoter region of the serotonin transporter gene serotonin-transporter-linked polymorphic region (5HTTLPR), has been suggested to affect serotonin uptake and may play a role in the pathogenesis of depression, obsessive-compulsive disorder (OCD) and aggressive behavior.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert Schwamborn et al.
Journal of neurochemistry, 137(3), 423-435 (2016-03-01)
The serotonin transporter (SERT) is responsible for high-affinity serotonin (5-HT) uptake from extracellular fluid and is a prominent pharmacological target in the treatment of depression. In recent years, depression has also been linked to immune system activation. Inflammatory conditions can
Mahesh Mundalil Vasu et al.
PloS one, 11(10), e0164425-e0164425 (2016-10-08)
Evidence suggests neuroprotective effects of fluoxetine, a selective serotonin reuptake inhibitor (SSRI), on the developed neurons in the adult brain. In contrast, the drug may be deleterious to immature or undifferentiated neural cells, although the mechanism is unclear. Recent investigations
Fluoxetine Increases the Expression of miR-572 and miR-663a in Human Neuroblastoma Cell Lines
Mundalil Vasu M
PLoS ONE, 11 (2016)
Allelic heterogeneity at the serotonin transporter locus (SLC6A4) confers susceptibility to autism and rigid-compulsive behaviors.
Sutcliffe JS
American Journal of Human Genetics, 77, 265-279 (2005)
Mariko Nakamura et al.
Human psychopharmacology, 37(2), e2818-e2818 (2021-09-21)
The aim of this study was evaluation of the association between severity of pain and expression of total or ubiquitinated serotonin transporter (SERT) protein in patients with burning mouth syndrome and atypical odontalgia (BMS/AO), who were treated by duloxetine. Patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service