Skip to Content
Merck
All Photos(1)

Key Documents

AB1594P

Sigma-Aldrich

Anti-Serotonin Transporter Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

Serotonin Transporter (SERT). An antibody raised against a similar peptide has been shown to react with a SERT in baculovirus cells expressing recombinant rat and human SERT. Antibody activity may be reduced considerably when detecting endogenous SERT from rat or human cells. SERT has been observed to migrate as 94 kDa (rat platelet), 80 kDa (pulmonary membranes) and 76 kDa (rat brain). This could be due to differences in postranslational membrane modifications (e.g. glycosylation).

Immunogen

A 15 amino acid sequence between transmembrane domain 7 and 8 of the rat serotonin transporter. This region is presumably from the extracellular domain.

Application

Anti-Serotonin Transporter Antibody is an antibody against Serotonin Transporter for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Neuronal & Glial Markers
Western blot: 1-10 μg/mL


Cell pellets are resuspended in 100 μL lysis buffer (25 mM Tris, pH 7.6, 150 mM NaCl, 1 μg/mL aprotonin, 10 μg/mL leupeptin, 1 mM phenylmethylsulfonylfluoride, 1 mM sodium orthovanadate, 1 mM EDTA, and 1% Triton X-100). Samples were incubated for 1 hour at 4°C and were pelleted at high speed for 10 minutes at 4°C, and 100 μg protein per sample was subjected to 10% sodium dodecyl sulfate-polyacrylamide gel electrophoresis.





ELISA: 1:100,000 primary EIA using immunogen only (50-100 ng SERT peptide (Catalog Number AG297)/well). Not tested on purified protein at the stated dilutions.


Optimal working dilutions must be determined by end user.

Physical form

Affinity purified immunoglobulin. Liquid in 100 mM Tris, pH 7.5 with 0.2% BSA and 0.05% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christophe Wersinger et al.
The European journal of neuroscience, 24(1), 55-64 (2006-08-03)
alpha-Synuclein (alpha-Syn), a protein primarily localized in the presynaptic compartment of neurons, is known to regulate dopaminergic neurotransmission by negatively modulating dopamine transporter activity and regulating its trafficking to or away from the cell surface. Given the considerable homology between
A M Thompson et al.
Developmental neuroscience, 27(1), 1-12 (2005-05-12)
To investigate the putative role of serotonin (5-HT) in auditory brainstem development, the expression of the 5-HT transporter (5-HTT) was evaluated in the normal mouse brainstem at 6 different postnatal ages. The brains of C3H/HeJ mice at birth (P0) and
Christophe Wersinger et al.
Neuroscience letters, 453(3), 157-161 (2009-05-12)
Human alpha-synuclein (alpha-Syn) is instrumental in maintaining homeostasis of monoamine neurotransmitters in brain, through its trafficking, and regulation of the cell surface expression and, thereby, activity of dopamine, serotonin and norepinephrine transporters. Here we have investigated whether other members of
V M Pickel et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 19(17), 7356-7366 (1999-08-25)
Extracellular levels of serotonin [5-hydroxytryptamine (5-HT)] in the nucleus accumbens (NAc) can influence both cognitive and motor functions involving extensive connections with the frontal cortex. The 5-HT levels reflect vesicular release and plasmalemmal reuptake through the serotonin transporter (SERT). We
Anne Katrine Z Johansen et al.
Pulmonary circulation, 6(1), 82-92 (2016-05-11)
Pulmonary arterial hypertension (PAH) is a devastating vasculopathy that predominates in women and has been associated with dysregulated estrogen and serotonin signaling. Overexpression of the serotonin transporter (SERT(+)) in mice results in an estrogen-dependent development of pulmonary hypertension (PH). Estrogen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service