Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500950

Sigma-Aldrich

Anti-SLC6A4/5HTT/SERT antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HGNC:11050, Anti-SERT

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC6A4(6532)

General description

Solute carrier family 6 member 4 (SLC6A4), also known as 5-hydroxytryptamine transporter (5-HTT), is an integral membrane protein, encoded by the gene mapped to human chromosome 17q11.2. The encoded protein is a member of SLC6 gene family and is mainly expressed in brain and blood cells.

Immunogen

Peptide with sequence C-LHQGERETWGK from the internal region of the protein sequence according to NP_001036.1.

Application

Anti-SLC6A4/5HTT/SERT antibody produced in goat has been used in immunolabelling and Western blotting.

Biochem/physiol Actions

Solute carrier family 6 member 4 (SLC6A4) catalyzes the transport of neurotransmitter serotonin from the synaptic space into presynaptic neurons. It also plays a vital role in mood and anxiety regulation. Mutation on the gene might alter the expression of serotonin (5HT) in the synaptic cleft, which ultimately leads to schizophrenia. Allelic variations in the gene increase the risk of susceptibility to autism and rigid-compulsive behaviors.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linkage mapping of serotonin transporter protein gene SLC6A4
on chromosome 17
J. Gelernter
Human Genetics (2005)
Allelic Heterogeneity at the Serotonin Transporter Locus (SLC6A4) Confers Susceptibility to Autism and Rigid-Compulsive Behaviors
James S
Academic Journal of Interdisciplinary Studies (2005)
miR-15a and miR-16 regulate serotonin transporter expression in human placental and rat brain raphe cells
Pablo R
The International Journal of Neuropsychopharmacology (2013)
Pablo R Moya et al.
The international journal of neuropsychopharmacology, 16(3), 621-629 (2012-05-09)
The serotonin transporter (SERT) is a key regulatory molecule in serotonergic transmission implicated in numerous biological processes relevant to human disorders. Recently, it was shown that SERT expression is controlled by miR-16 in mouse brain. Here, we show that SERT
Nadia Postupna et al.
The American journal of pathology, 187(4), 884-895 (2017-02-19)
Cognitive impairment in older individuals is a complex trait that in population-based studies most commonly derives from an individually varying mixture of Alzheimer disease, Lewy body disease, and vascular brain injury. We investigated the molecular composition of synaptic particles from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service