Skip to Content
Merck
All Photos(3)

Documents

SAB2500963

Sigma-Aldrich

Anti-Smooth Muscle α-Actin antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ACTA2, Anti-OTTHUMP00000020042

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

technique(s)

ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTA2(59)

General description

Actin, a highly conserved protein, is a major component of both the cytoskeletal and contractile structures in all cell types. It varies in amount related to the type of differentiation and to the functional state of cells and tissues. Actin can be found in two different forms of aggregation, the globular or the fibrillar state, and at least six distinct isoforms occur in vertebrates. The actins exhibit over 90% sequence homology, but each isoform has a unique NH2-terminal sequence. The isoforms are comprised of three α actins (skeletal, cardiac, smooth), one β actin (β-non-muscle) and two γ actins (γ smooth muscle and γ non-muscle). Difficulties have been encountered in producing polyclonal antibodies due to the highly conserved nature of actin.
Goat polyclonal anti-smooth muscle α-Actin antibody reacts with canine, human, mouse, and rat smooth muscle α-actins (ACTA2).

Immunogen

Peptide with sequence EEEDSTALVC from the N Terminus of the protein sequence according to NP_001604.1.

Application

Goat polyclonal anti-smooth muscle α-Actin antibody is used to tag smooth muscle α-actin (ACTA2) for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of smooth muscle α-actin in smooth muscle structure and as a marker of myofibroblast formation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Riccardo Coletta et al.
Journal of tissue engineering and regenerative medicine, 10(2), 178-182 (2015-08-04)
The ability to grow embryonic organs ex vivo provides an opportunity to follow their differentiation in a controlled environment, with resulting insights into normal development. Additionally, similar strategies can be used to assess effects on organogenesis of physical and chemical
Rebecca M Shaffer et al.
American journal of obstetrics and gynecology, 221(4), 330-330 (2019-05-19)
The use of polypropylene prolapse mesh to treat pelvic organ prolapse has been limited by mesh-related complications. Gynemesh PS mesh, implanted via sacrocolpopexy in rhesus macaques, had a negative impact on the vagina with thinning of vaginal muscularis and decreased
Mayur J Patil et al.
eNeuro, 10(12) (2023-11-22)
Transient receptor potential ankyrin 1 (TRPA1) is a polymodal cation channel that is activated by electrophilic irritants, oxidative stress, cold temperature, and GPCR signaling. TRPA1 expression has been primarily identified in subsets of nociceptive sensory afferents and is considered a
Xi Zeng et al.
Experimental and therapeutic medicine, 15(1), 1062-1068 (2018-02-13)
The aim of the present study was to elucidate the effect of transmembrane protein 16A (TMEM16A) on portal vein smooth muscle cell (PVSMC) proliferation associated with portal vein remodeling in portal hypertension (PHT). Sprague-Dawley rats were subjected to bile duct
Mitchell R Ladd et al.
Tissue engineering. Part C, Methods, 24(6), 331-345 (2018-04-12)
Short bowel syndrome (SBS) is a major cause of morbidity and mortality in the pediatric population, for which treatment options are limited. To develop novel approaches for the treatment of SBS, we now focus on the development of a tissue-engineered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service