Skip to Content
Merck
All Photos(3)

Key Documents

M4276

Sigma-Aldrich

Anti-MYH-1 Antibody

mouse monoclonal, MY-32

Synonym(s):

Anti-Myosin Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Monoclonal Anti-Myosin (Skeletal, Fast) antibody produced in mouse, clone MY-32, ascites fluid

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

MY-32, monoclonal

contains

15 mM sodium azide

species reactivity

rat, chicken, rabbit, mouse, human, bovine, guinea pig, feline

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:400 using skeletal muscle tissue
indirect immunofluorescence: 1:400
western blot: 1:1,000 using rabbit leg muscle extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Localizes an epitope on the myosin heavy chain. Stains the fast (type II) and neonatal isomyosin molecules found in skeletal muscle, but does not stain cardiac muscle, smooth muscle or non-muscle myosin in cultured cells. Does react with human rhabdomyosarcomas.
Monoclonal Anti-Skeletal Myosin (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Myosin is a 480,000 dalton protein known to interact with actin in muscle and in non-muscle cells. It contains two identical heavy chains (200,000 daltons each) and four light chains (15,000-26,000 daltons). Myosin molecules consist of two major regions: tails (rods) and heads; they aggregate into filaments through the tail region and interact with actin and with ATP through the head region. Multiple forms of myosin heavy chains exist for each muscle type-skeletal, cardiac, smooth and non-muscle isomyosin forms exist in different types of skeletal muscle, depending on the physiological function of the muscle. These are designated at type I (slow twitch) and type II (fast-twitch). Type II fibers can be further subdivided in types IIA, IIB, and IIC.

Specificity

Monoclonal Anti-Skeletal Myosin is specific for the myosin heavy chain. It does not stain human or animal cardiac or smooth muscle myosin or cells grown by tissue culture (nonmuscle myosin). It has been demonstrated on human skeletal muscle that the antibody stains the fast twitch (type II) isomyosin molecules. Monoclonal Anti-Skeletal Myosin antibody does react with human rhabdomyosarcomas.

Immunogen

rabbit muscle myosin.

Application

Monoclonal Anti-Myosin (Skeletal, Fast) antibody produced in mouse has been used in:
  • immunohistochemistry
  • immunostaining
  • western blotting at a dilution 1:1000 and 1:90000†
  • indirect immunofluorescence (dilution 1:400) of formalin-fixed, paraffin-embedded sections of human or animal skeletal muscle tissue preparation.
  • dot immunobinding on muscle extracts or purified myosin preparations
Monoclonal Anti-Skeletal Myosin may be used for staining of human, rabbit, rat, mouse, bovine, chicken and guinea pig skeletal myosin. Monoclonal Anti-Skeletal Myosin antibody to fast-twitch skeletal myosin may be used for detecting cross striated muscle differentiation in tumors. The antibody localizes an epitope on the myosin chain that is stable to the routine formalin-fixation and paraffin-embedding process.
The level of mysosin (fast) in serum samples from sportsmen with past injury was determined by western blot using monoclonal mouse anti-myosin (skeletal/fast) as the primary antibody at a dilution of 1:90000.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Contractility and kinetics of human fetal and human adult skeletal muscle
Racca AW, et al.
The Journal of Physiology, 591(12), 3049-3061 (2013)
David Vaughan et al.
PloS one, 11(3), e0149046-e0149046 (2016-03-18)
A silencer region (I-allele) within intron 16 of the gene for the regulator of vascular perfusion, angiotensin-converting enzyme (ACE), is implicated in phenotypic variation of aerobic fitness and the development of type II diabetes. We hypothesised that the reportedly lower
An optimized histochemical method to assess skeletal muscle glycogen and lipid stores reveals two metabolically distinct populations of type I muscle fibers
Prats C, et al.
PLoS ONE, 8(10), e77774-e77774 (2013)
Shanshan Wang et al.
Frontiers in cell and developmental biology, 8, 618617-618617 (2021-01-30)
Enhancer of zeste homolog 2 (EZH2) is the catalytic subunit of polycomb repressive complex 2 and contains a SET domain that catalyzes histone H3 trimethylation on lysine 27 (H3K27me3) to generate an epigenetic silencing mark. EZH2 interacts with transcription factors
S Cefalù et al.
Cell cycle (Georgetown, Tex.), 14(6), 894-901 (2015-03-20)
p53 family members, p63 and p73, play a role in controlling early stage of myogenic differentiation. We demonstrated that TAp63gamma, unlike the other p53 family members, is markedly up-regulated during myogenic differentiation in murine C2C7 cell line. We also found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service