Skip to Content
Merck
All Photos(6)

Key Documents

A2172

Sigma-Aldrich

Anti-α-Actin-1 (ACTA1) antibody

mouse monoclonal, 5C5

Synonym(s):

Mouse Anti-Actin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Monoclonal Anti-Actin (α-Sarcomeric) antibody produced in mouse, clone 5C5, ascites fluid

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

5C5, monoclonal

contains

15 mM sodium azide

species reactivity

rabbit, carp, rat, sheep, guinea pig, human, snake, bovine, frog

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:500 using human skeletal and cardiac muscle
immunohistochemistry (frozen sections): suitable using human skeletal and cardiac muscle
indirect ELISA: suitable
western blot: suitable

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Monoclonal Anti-α-Sarcomeric Actin (mouse IgM isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Actin is one of the most conserved eukaryotic proteins and the isoforms show >90% overall sequence homology. It is expressed in mammals and birds as six isoforms. Four of the six represent differentiation markers of muscle tissues. The other two are found in practically all cells.

Immunogen

rabbit striated muscle actin.

Application

Monoclonal Anti-Actin (α-Sarcomeric) antibody produced in mouse has also been used
  • immunofluorescence
  • double immunostaining
  • double labelling terminal deoxynucleotidyl transferase dUTP nick end labeling (TUNEL) immunofluorescence staining.

Monoclonal mouse anti-actin antibody was used as a loading control for western blot analysis of immunoprecipitated proteins from rat dorsal root ganglion cocultures.
Mouse monoclonal anti-actin (alpha-sarcomeric) antibody has been used for western blot, and immunohistochemistry applications.

Biochem/physiol Actions

Actin is a cytoskeletal protein that regulates cell motility, secretion, phagocytosis and cytokinesis. The NH2-terminal of actin may function as antigen. This terminal may also modulate actin interactions and may associate with proteins such as myosin.

Physical form

The product is provided as ascites fluid containing 0.1% sodium azide as a preservative.

Other Notes

To view an Actin antibody selection guide, please visit www.sigmaaldrich.com/actin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PEDF and PEDF-derived peptide 44mer stimulate cardiac triglyceride degradation via ATGL
Zhang H, et al.
Journal of Translational Medicine, 13(1), 68-68 (2015)
PEDF improves cardiac function in rats subjected to myocardial ischemia/reperfusion injury by inhibiting ROS generation via PEDF-R
Zhao Q, et al.
International Journal of Molecular Medicine, 41(6), 3243-3252 (2018)
Monoclonal antibodies against muscle actin isoforms: epitope identification and analysis of isoform expression by immunoblot and immunostaining in normal and regenerating skeletal muscle
Chaponnier C and Gabbiani G
F1000Research, 5 (2016)
Forced myocardin expression enhances the therapeutic effect of human mesenchymal stem cells after transplantation in ischemic mouse hearts
Grauss RW, et al.
Stem Cells, 26(4), 1083-1093 (2008)
Sarah A Swager et al.
Cardiovascular pathology : the official journal of the Society for Cardiovascular Pathology, 24(3), 160-167 (2014-12-03)
Claudin-5 is transcriptionally downregulated resulting in dramatically reduced protein levels in human heart failure. Studies in mice have demonstrated that reduced claudin-5 levels occur prior to cardiac damage and far before reduced whole heart function. Therefore, claudin-5 may be a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service