Skip to Content
Merck
All Photos(3)

Key Documents

SAB2104562

Sigma-Aldrich

Anti-RBM47 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp686F02235, Anti-FLJ20273, Anti-FLJ21344, Anti-FLJ21643

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

57 kDa

species reactivity

mouse, guinea pig, rat, human, dog, horse, bovine, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RBM47(54502)

Immunogen

Synthetic peptide directed towards the middle region of human RBM47

Application

Anti-RBM47 antibody produced in rabbit has been used in Western blotting.

Sequence

Synthetic peptide located within the following region: HFTSREDAVHAMNNLNGTELEGSCLEVTLAKPVDKEQYSRYQKAARGGGA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annabel N Smith et al.
Journal of bioenergetics and biomembranes, 40(4), 371-380 (2008-08-30)
The multi-subunit vacuolar-type H(+)-ATPase consists of a V(1) domain (A-H subunits) catalyzing ATP hydrolysis and a V(0) domain (a, c, c', c", d, e) responsible for H(+) translocation. The mammalian V(0) d subunit is one of the least-well characterized, and
Yueqin Yang et al.
Molecular and cellular biology, 36(11), 1704-1719 (2016-04-06)
The epithelial-to-mesenchymal transition (EMT) is an essential biological process during embryonic development that is also implicated in cancer metastasis. While the transcriptional regulation of EMT has been well studied, the role of alternative splicing (AS) regulation in EMT remains relatively
Veronica Ruta et al.
Cell reports. Medicine, 5(2), 101411-101411 (2024-02-08)
Pancreatic ductal adenocarcinoma (PDAC) is characterized by extremely poor prognosis. PDAC presents with molecularly distinct subtypes, with the basal-like one being associated with enhanced chemoresistance. Splicing dysregulation contributes to PDAC; however, its involvement in subtype specification remains elusive. Herein, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service