Skip to Content
Merck
All Photos(8)

Key Documents

HPA006347

Sigma-Aldrich

Anti-RBM47 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-RNA-binding motif protein 47, Anti-RNA-binding protein 47

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

HAMNNLNGTELEGSCLEVTLAKPVDKEQYSRYQKAARGGGAAEAAQQPSYVYSCDPYTLAYYGYPYNALIGPNRDYFVKAGSIRGRGRGAAGNRAPGPRGSYLGGYSAGRGIYSRYHEGKGKQQEKGYELVPNLEIPTVN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RBM47(54502)

General description

RBM47 (RNA binding motif protein 47) is an mRNA-interacting protein. It is composed of three classical RNA recognition motifs (RRM domains). It is a homolog of the RNA-editing proteins Apobec1 complementation factor (A1CF) and hnRNP-Q.

Immunogen

RNA-binding protein 47 recombinant protein epitope signature tag (PrEST)

Application

Anti-RBM47 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

RBM47 (RNA binding motif protein 47) suppresses the breakdown rate of mRNA. This causes accumulation of protein inside cells, leading to reduced tumor growth. It is under-expressed in metastatic tumor, and has been identified as a breast cancer tumor suppressor.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86761

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Sakurai et al.
Oncogene, 35(38), 5000-5009 (2016-03-01)
RNA-binding proteins provide a new layer of posttranscriptional regulation of RNA during cancer progression. We identified RNA-binding motif protein 47 (RBM47) as a target gene of transforming growth factor (TGF)-β in mammary gland epithelial cells (NMuMG cells) that have undergone
Ruilei Li et al.
Oncology letters, 19(4), 3111-3122 (2020-03-29)
Lung cancer is the leading cause of cancer-associated mortality worldwide. In China, in particular, lung cancer mortality has markedly increased and is likely to continue to rise. RNA-binding proteins are pivotal to the development and progression of a variety of
Sakari Vanharanta et al.
eLife, 3, doi:10-doi:10 (2014-06-06)
The mechanisms through which cancer cells lock in altered transcriptional programs in support of metastasis remain largely unknown. Through integrative analysis of clinical breast cancer gene expression datasets, cell line models of breast cancer progression, and mutation data from cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service