Skip to Content
Merck
All Photos(1)

Key Documents

S9701

Sigma-Aldrich

Sulbactam

Synonym(s):

(2S,5R)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 4,4-dioxide, Betamaze, CP 45899, CP-45,899, Penicillanic acid 1,1-dioxide, Penicillanic acid S,S-dioxide, Penicillanic acid dioxide, Penicillanic acid sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO5S
CAS Number:
Molecular Weight:
233.24
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D ≥+225°, c = 1 in H2O

color

white to tan

solubility

H2O: ≥18 mg/mL

storage temp.

2-8°C

SMILES string

CC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O

InChI

1S/C8H11NO5S/c1-8(2)6(7(11)12)9-4(10)3-5(9)15(8,13)14/h5-6H,3H2,1-2H3,(H,11,12)/t5-,6+/m1/s1

InChI key

FKENQMMABCRJMK-RITPCOANSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulbactam is a semi-synthetic penicillinate sulfone derived from 6-aminopenicllanic acid. This β-lactam compound is characterized by a β-lactam ring.

Application

Sulbactam has been used as a β-Lactamase inhibitor in β-lactamase inhibitor assay. It has also been used to evaluate its antimicrobial pharmacodynamic effects against extremely drug-resistant Acinetobacter baumannii. Sulbactam may be used in cell signaling studies.

Biochem/physiol Actions

Sulbactam acts as an irreversible inhibitor of β-lactamases that inactivate β-lactams such as penicillin and cephalosporin. It is also active against bacteroides and certain chromosomally mediated enzymes of Gram-negative bacteria. It exhibits limited antimicrobial activity. However, sulbactam in combination with other potential antibiotics exhibits therapeutic effects against multidrug-resistant Acinetobacter baumannii infections.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mallika Sengupta et al.
Cureus, 14(2), e21802-e21802 (2022-03-08)
Background Acinetobacter species are known to be important hospital-acquired pathogens. Unfortunately, multidrug-resistant Acinetobacter spp. has very limited options for an effective treatment. Aim To identify the common pathogens causing lower respiratory tract infections (LRTI), their antimicrobial susceptibility pattern, and determine the minimum inhibitory
Sarah M Drawz et al.
Clinical microbiology reviews, 23(1), 160-201 (2010-01-13)
Since the introduction of penicillin, beta-lactam antibiotics have been the antimicrobial agents of choice. Unfortunately, the efficacy of these life-saving antibiotics is significantly threatened by bacterial beta-lactamases. beta-Lactamases are now responsible for resistance to penicillins, extended-spectrum cephalosporins, monobactams, and carbapenems.
G S Singh
Mini reviews in medicinal chemistry, 4(1), 93-109 (2004-02-03)
Beta-lactam ring-containing compounds such as penicillins, ampicillin, amoxicillin, cephalosporins and carbapenems are among the most famous antibiotics. This article reviews the recent developments in study of cephems, oxacephems, penams and sulbactam. Many of the compounds reviewed have potential antibacterial activity
Krisztina M Papp-Wallace et al.
Antimicrobial agents and chemotherapy, 56(11), 5687-5692 (2012-08-22)
Acinetobacter baumannii is an increasingly problematic pathogen in United States hospitals. Antibiotics that can treat A. baumannii are becoming more limited. Little is known about the contributions of penicillin binding proteins (PBPs), the target of β-lactam antibiotics, to β-lactam-sulbactam susceptibility
Bahadir M Gulluoglu et al.
Annals of surgery, 257(1), 37-43 (2012-09-25)
To assess the impact of prophylactic antibiotics on the prevention of surgical site infection (SSI) and the cost-effectiveness of this prophylaxis for breast cancer surgery in overweight or obese women. SSI is higher than expected after breast surgery. Obesity was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service