Skip to Content
Merck
All Photos(2)

Key Documents

S9666

Sigma-Aldrich

Sphingosine 1-phosphate

≥95% (TLC), powder, lipid second messenger

Synonym(s):

(2S,3R,4E)-2-Amino-4-octadecene-1,3-diol 1-phosphate, D-erythro-Sphingosine 1-phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H38NO5P
CAS Number:
Molecular Weight:
379.47
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Sphingosine 1-phosphate, ≥95%, powder

Quality Level

Assay

≥95%

form

powder

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC\C=C\[C@@H](O)[C@@H](N)COP(O)(O)=O

InChI

1S/C18H38NO5P/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(20)17(19)16-24-25(21,22)23/h14-15,17-18,20H,2-13,16,19H2,1H3,(H2,21,22,23)/b15-14+/t17-,18+/m0/s1

InChI key

DUYSYHSSBDVJSM-KRWOKUGFSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A lipid second messenger that binds to S1P1 and S1P3 receptors. Mobilizes intracellular Ca2+ stores and decreases cellular cAMP. Activates phospholipase D. S1P stimulates the migration of endothelial cells but inhibits the migration of other cell types. Induces angiogenesis.

Features and Benefits

This compound is featured on the Lysophospholipid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaiqi Sun et al.
Nature communications, 7, 12086-12086 (2016-07-16)
Sphingosine-1-phosphate (S1P) is a bioactive signalling lipid highly enriched in mature erythrocytes, with unknown functions pertaining to erythrocyte physiology. Here by employing nonbiased high-throughput metabolomic profiling, we show that erythrocyte S1P levels rapidly increase in 21 healthy lowland volunteers at
H Okamoto et al.
Molecular and cellular biology, 20(24), 9247-9261 (2000-11-30)
Sphingosine-1-phosphate (S1P) is a bioactive lysophospholipid that induces a variety of biological responses in diverse cell types. Many, if not all, of these responses are mediated by members of the EDG (endothelial differentiation gene) family G protein-coupled receptors EDG1, EDG3
William S Szczepaniak et al.
Translational research : the journal of laboratory and clinical medicine, 152(5), 213-224 (2008-11-18)
S1P has been demonstrated to protect against the formation of lipopolysaccharide (LPS)-induced lung edema when administered concomitantly with LPS. In the current study, we sought to determine the effectiveness of S1P to attenuate lung injury in a translationally relevant canine
Roland Martin et al.
Current topics in microbiology and immunology, 378, 149-170 (2014-04-15)
The development of fingolimod, an unselective functional antagonist of the interactions between sphingosine 1 phosphate (S1P) and sphingosine 1 phosphate receptors (S1PRs), as the first oral therapy for multiple sclerosis (MS) has been a milestone. The parallel intensive research on
Tsuyoshi Nishi et al.
Biochimica et biophysica acta, 1841(5), 759-765 (2013-08-08)
Sphingosine 1-phosphate (S1P) is a lipid mediator that plays important roles in diverse cellular functions such as cell proliferation, differentiation and migration. S1P is synthesized inside the cells and subsequently released to the extracellular space, where it binds to specific

Questions

  1. Will using methanol as a solvent to dissolve the powder affect or disrupt the huvect cell line?

    1 answer
    1. Another supplier (APExBIO) suggests using a solvent of 0.3M NaOH for in vitro work, as the substance is soluble in it at less than 4 mg/mL. They also recommend warming the tube at 37°C and shaking it in an ultrasonic bath for a while to achieve higher solubility. The stock solution can be stored below -20°C for several months.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service