Skip to Content
Merck
All Photos(1)

Key Documents

860659P

Avanti

sphingosine-1-phosphate-d7

Avanti Research - A Croda Brand 860659P, powder

Synonym(s):

D-erythro-sphingosine-d7-1-phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H31D7NO5P
CAS Number:
Molecular Weight:
386.51
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860659P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860659P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]([C@]([NH3+])([H])COP([O-])(O)=O)(O)/C=C/CCCCCCCCCCC(C(C([2H])([2H])[2H])([2H])[2H])([2H])[2H]

InChI

1S/C18H38NO5P/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(20)17(19)16-24-25(21,22)23/h14-15,17-18,20H,2-13,16,19H2,1H3,(H2,21,22,23)/b15-14+/t17-,18+/m0/s1/i1D3,2D2,3D2

InChI key

DUYSYHSSBDVJSM-AVMMVCTPSA-N

General description

Sphingosine-1-phosphate (S1P) or D-erythro-sphingosine-d7-1-phosphate, is a deuterated sphingolipid. S1P, a zwitterionic sphingolipid, is an important cellular metabolite and signaling lipid in animal cells. It has also been found in insects, yeasts and plants. S1P is synthesized from the sphingosine kinase (Sph) 1 and SphK2 present in cytosol and nucleus, respectively.

Application

Sphingosine-1-phosphate (S1P)-d7 has been used as a standard in ultra-high performance liquid chromatography (UHPLC)-Q exactive for validation and quantitative analyses of S1P.

Biochem/physiol Actions

Sphingosine-1-phosphate (S1P) plays a vital role in mammalian brain and cardiac development. It is essential for maturation of the systemic circulatory system and lymph vessels. Additionally, S1P is implicated in trafficking and effector functions of lymphocytes and other blood stem cells. It also provides protection against various forms of tissue injury. S1P also acts as a tumor promoting lipid. S1P has an ability to enhance erythrocyte glycolysis and oxygen delivery therefore, it can be used to treat tissue hypoxia.

Packaging

5 mL Amber Glass Screw Cap Vial (860659P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An update on sphingosine-1-phosphate and other sphingolipid mediators
Fyrst H and Saba JD
Nature Chemical Biology, 6(7), 489-489 (2010)
Yugesh Kharel et al.
The Biochemical journal, 477(5), 925-935 (2020-02-18)
The interplay of sphingosine 1-phosphate (S1P) synthetic and degradative enzymes as well as S1P exporters creates concentration gradients that are a fundamental to S1P biology. Extracellular S1P levels, such as in blood and lymph, are high relative to cellular S1P.
Structural and functional insight of sphingosine 1-phosphate-mediated pathogenic metabolic reprogramming in sickle cell disease
Sun K, et al.
Scientific reports, 7(1), 15281-15281 (2017)
Sphingosine-1-phosphate promotes erythrocyte glycolysis and oxygen release for adaptation to high-altitude hypoxia
Sun K, et al.
Nature Communications, 7(7), 12086-12086 (2016)
Iulia Zoicas et al.
Cells, 9(5) (2020-05-24)
Human and murine studies identified the lysosomal enzyme acid sphingomyelinase (ASM) as a target for antidepressant therapy and revealed its role in the pathophysiology of major depression. In this study, we generated a mouse model with overexpression of Asm (Asm-tgfb)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service