Skip to Content
Merck
All Photos(5)

Documents

S9129

Sigma-Aldrich

Somatostatin

≥97% (HPLC), powder

Synonym(s):

Growth hormone release inhibiting factor, SRIF, Somatostatin-14, Somatotropin release inhibiting factor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C76H104N18O19S2
CAS Number:
Molecular Weight:
1637.88
Beilstein:
6436064
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥97% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

[H]N1[C@@H](Cc2ccccc2)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](Cc4c[nH]c5ccccc45)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](Cc6ccccc6)C(=O)N[C@@H]([C@@H](C)O)C(=O)NC(CO)C(=O)N[C@@H](CSSC[C@H](NC(=O)CNC(=O)[C@H](C)N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(N)=O)C1=O)C(O)=O

InChI

1S/C76H104N18O19S2/c1-41(79)64(100)82-37-61(99)83-58-39-114-115-40-59(76(112)113)92-72(108)57(38-95)91-75(111)63(43(3)97)94-71(107)54(33-46-23-11-6-12-24-46)90-74(110)62(42(2)96)93-66(102)51(28-16-18-30-78)84-69(105)55(34-47-36-81-49-26-14-13-25-48(47)49)88-68(104)53(32-45-21-9-5-10-22-45)86-67(103)52(31-44-19-7-4-8-20-44)87-70(106)56(35-60(80)98)89-65(101)50(85-73(58)109)27-15-17-29-77/h4-14,19-26,36,41-43,50-59,62-63,81,95-97H,15-18,27-35,37-40,77-79H2,1-3H3,(H2,80,98)(H,82,100)(H,83,99)(H,84,105)(H,85,109)(H,86,103)(H,87,106)(H,88,104)(H,89,101)(H,90,110)(H,91,111)(H,92,108)(H,93,102)(H,94,107)(H,112,113)/t41-,42+,43+,50-,51-,52-,53-,54-,55-,56-,57?,58-,59-,62-,63-/m0/s1

InChI key

NHXLMOGPVYXJNR-FQSIDJEASA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Ala-Gly-Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Thr-Ser-Cys

General description

Somatostatin (SST) is a peptide.
Somatostatin is expressed in endocrine cells and nerve fibers of the gastrointestinal tract. It is also expressed in other tissues.

Application

Somatostatin has been used:
  • as a standard antigen to detect antibodies in an indirect enzyme-linked immunosorbent assay (ELISA) method
  • to study the effect of somatostatin on oxytocin release during pregnancy in rats
  • to test the specificity of the avidin-biotinylated horseradish peroxidase immunocytochemical procedure to visualize somatostatin-immunoreactive (SS-IR) neurons

Biochem/physiol Actions

Somatostatin (SST) performs several functions like anti-proliferation, pro-apoptosis and anti-migration. Somatostatin performs physiological activities with the help of various G protein-coupled receptor subtypes (SST receptor (SSTR)-1, SSTR2A, SSTR2B, and SSTR3-5).
Somatostatin regulates the secretion of hormones and bioactive peptides. It acts as an inhibitor in the digestive tract by inhibiting gastrin release. In the pancreatic islets, it inhibits glucagon and insulin release. Somatostatin and its synthetic analogs have been used in the treatment of neuroendocrine tumors.

Other Notes

Lyophilized from 0.1% TFA in H2O

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

No loss of somatostatin-immunoreactive neurons in the hippocampal dentate hilus of alcohol-withdrawal-kindled rats.
Ulrichsen J
Alcohol and Alcoholism (Oxford, Oxfordshire), 31(4), 411-419 (1996)
Somatostatin inhibits insulin and glucagon secretion via two receptors subtypes: an in vitro study of pancreatic islets from somatostatin receptor 2 knockout mice.
Strowski MZ
Endocrinology, 141(1), 111-117 (2000)
Long-term effects of somatostatin 14 on the pituitary-ovarian axis in rats.
NestoroviA?, et al.
General Physiology and Biophysics, 33, 157-168 (2014)
Physiological role of somatostatin in the digestive tract: gastric acid secretion, intestinal absorption, and motility.
Krejs GJ
Scandinavian Journal of Gastroenterology. Supplement, 119, 47-53 (1986)
Construction and evaluation of the eukaryotic expression plasmid encoding two copies of somatostatin genes fused with hepatitis B surface antigen gene S.
Liang A
Vaccine, 26(23), 2935-2941 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service