Skip to Content
Merck
All Photos(1)

Key Documents

S0945000

Somatostatin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Growth hormone release inhibiting factor, SRIF, Somatostatin-14, Somatotropin release inhibiting factor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C76H104N18O19S2
CAS Number:
Molecular Weight:
1637.88
Beilstein:
6436064
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

somatostatin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[H]N1[C@@H](Cc2ccccc2)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](Cc4c[nH]c5ccccc45)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](Cc6ccccc6)C(=O)N[C@@H]([C@@H](C)O)C(=O)NC(CO)C(=O)N[C@@H](CSSC[C@H](NC(=O)CNC(=O)[C@H](C)N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(N)=O)C1=O)C(O)=O

InChI

1S/C76H104N18O19S2/c1-41(79)64(100)82-37-61(99)83-58-39-114-115-40-59(76(112)113)92-72(108)57(38-95)91-75(111)63(43(3)97)94-71(107)54(33-46-23-11-6-12-24-46)90-74(110)62(42(2)96)93-66(102)51(28-16-18-30-78)84-69(105)55(34-47-36-81-49-26-14-13-25-48(47)49)88-68(104)53(32-45-21-9-5-10-22-45)86-67(103)52(31-44-19-7-4-8-20-44)87-70(106)56(35-60(80)98)89-65(101)50(85-73(58)109)27-15-17-29-77/h4-14,19-26,36,41-43,50-59,62-63,81,95-97H,15-18,27-35,37-40,77-79H2,1-3H3,(H2,80,98)(H,82,100)(H,83,99)(H,84,105)(H,85,109)(H,86,103)(H,87,106)(H,88,104)(H,89,101)(H,90,110)(H,91,111)(H,92,108)(H,93,102)(H,94,107)(H,112,113)/t41-,42+,43+,50-,51-,52-,53-,54-,55-,56-,57?,58-,59-,62-,63-/m0/s1

InChI key

NHXLMOGPVYXJNR-FQSIDJEASA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Ala-Gly-Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Thr-Ser-Cys

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Somatostatin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Henrik H Hansen et al.
The Journal of pharmacology and experimental therapeutics, 350(3), 657-664 (2014-07-06)
Type 2 diabetes is characterized by impaired β-cell function associated with progressive reduction of insulin secretion and β-cell mass. Evidently, there is an unmet need for treatments with greater sustainability in β-cell protection and antidiabetic efficacy. Through an insulin and
Naoki Takada et al.
Nature communications, 5, 5333-5333 (2014-10-31)
The cellular diversity of interneurons in the neocortex is thought to reflect subtype-specific roles of cortical inhibition. Here we ask whether perturbations to two subtypes--parvalbumin-positive (PV+) and somatostatin-positive (SST+) interneurons--can be compensated for with respect to their contributions to cortical
Danh-Tai Hoang et al.
PloS one, 9(10), e110384-e110384 (2014-10-29)
Morphogenesis, spontaneous formation of organism structure, is essential for life. In the pancreas, endocrine α, β, and δ cells are clustered to form islets of Langerhans, the critical micro-organ for glucose homeostasis. The spatial organization of endocrine cells in islets
Amelie Soumier et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(9), 2252-2262 (2014-04-03)
Reduced expression of somatostatin (SST) is reported across chronic brain conditions including major depression and normal aging. SST is a signaling neuropeptide and marker of gamma-amino butyric acid (GABA) neurons, which specifically inhibit pyramidal neuron dendrites. Studies in auditory cortex
Lili Ren et al.
Cells, tissues, organs, 199(5-6), 329-341 (2014-12-05)
Human adipose tissue-derived stem cells (hASCs) are considered an ideal tool for the supply of insulin-producing cells to treat diabetes mellitus, with high differentiation efficiency. Islet neogenesis-associated protein (INGAP) is an initiator of islet neogenesis, and the peptide sequence comprising

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service