Skip to Content
Merck
All Photos(1)

Key Documents

S6883

Sigma-Aldrich

Substance P acetate salt hydrate

≥95% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C63H98N18O13S · xC2H4O2 · yH2O
CAS Number:
Molecular Weight:
1347.63 (anhydrous free base basis)
Beilstein:
5153552
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95% (HPLC)

form

powder

mol wt

1348 Da by calculation (Average Mass)

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

color

white

solubility

0.1 M acetic acid: 1 mg/mL

UniProt accession no.

storage temp.

−20°C

SMILES string

[H]O[H].CC(O)=O.CSCC[C@H](NC(=O)[C@H](CC(C)C)NC(=O)CNC(=O)[C@@H](Cc1ccccc1)NC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H]3CCCN3C(=O)[C@H](CCCCN)NC(=O)[C@@H]4CCCN4C(=O)[C@@H](N)CCCNC(N)=N)C(N)=O

InChI

1S/C63H98N18O13S.C2H4O2.H2O/c1-37(2)33-45(57(89)74-41(53(68)85)27-32-95-3)73-52(84)36-72-54(86)46(34-38-15-6-4-7-16-38)78-58(90)47(35-39-17-8-5-9-18-39)79-56(88)42(23-25-50(66)82)75-55(87)43(24-26-51(67)83)76-59(91)49-22-14-31-81(49)62(94)44(20-10-11-28-64)77-60(92)48-21-13-30-80(48)61(93)40(65)19-12-29-71-63(69)70;1-2(3)4;/h4-9,15-18,37,40-49H,10-14,19-36,64-65H2,1-3H3,(H2,66,82)(H2,67,83)(H2,68,85)(H,72,86)(H,73,84)(H,74,89)(H,75,87)(H,76,91)(H,77,92)(H,78,90)(H,79,88)(H4,69,70,71);1H3,(H,3,4);1H2/t40-,41-,42-,43-,44-,45-,46?,47-,48-,49-;;/m0../s1

InChI key

VHKFRLDSEZYWAC-CIYRQLMPSA-N

Gene Information

human ... TAC1(6863)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2

General description

Research area: Cell Signaling

Substance P is a member of the tachykinin family of peptides. It is encoded by the tachykinin precursor 1 (TAC1) gene mapped to human chromosome 7q21-q22. Substance P is a small peptide produced by macrophages, lymphocytes, eosinophils, dendritic cells and nerves. It is part of the neurokinin group.

Application

Substance P acetate salt hydrate has been:

  • used as a neurokinin 1 (NK1) receptor agonist for the activation of spinal nociceptive circuitry in mice
  • administered in central nucleus (ACE) in rat to study its positive reinforcing effects
  • used in competition binding assays
  • used as an inhibitor of myosin light chain kinase in isolated lymphatic vessels from rat

Biochem/physiol Actions

NK-1 agonist; potent vasodilator and hypotensive agent.
NK-1 agonist; potent vasodilator and hypotensive agent; induces salivation; increases capillary permeability; induces mast cell degranulation; putative neurotransmitter in sensory (pain) afferents.
Substance P has proinflammatory effects on epithelial and immune cells. It has been associated with inflammatory diseases related to gastrointestinal and respiratory systems. Substance P modulates contractility of smooth muscles, transport of ions in the epithelium, vascular permeability and immune functions in the gut.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of myosin light chain phosphorylation
Zawieja A, et al.
American Journal of Physiology. Heart and Circulatory Physiology, 297(1), H726-H734 (2009)
Tae-You Kim et al.
Cancer research, 66(15), 7490-7501 (2006-08-04)
Malignant glioma is the most common central nervous system tumor of adults and is associated with a significant degree of morbidity and mortality. Gliomas are highly invasive and respond poorly to conventional treatments. Gliomas, like other tumor types, arise from
The effect of capsaicin on substance P expression in pulp tissue inflammation.
Caviedes-Bucheli J
International Endodontic Journal, 38(1), 30-33 (2005)
G F Joos et al.
Allergy, 55(4), 321-337 (2000-04-27)
The sensory neuropeptides substance P (SP) and neurokinin A (NKA) are localized to sensory airway nerves, from which they can be released by a variety of stimuli, including allergen, ozone, or inflammatory mediators. Sensory nerves containing these peptides are relatively
The associations of TAC1 gene polymorphisms with major depressive disorder
Won E, et al
Molecular & Cellular Toxicology, 129-136 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service