Skip to Content
Merck
All Photos(1)

Key Documents

S7198

Sigma-Aldrich

Sivelestat sodium salt hydrate

≥98% (HPLC), solid

Synonym(s):

EI 546 sodium salt hydrate, Elaspol sodium salt hydrate, LY 544349 sodium salt hydrate, Ono-5046 sodium salt hydrate, o-(p-Hydroxybenzenesulfonamido)hippuric acid, pivalate (ester) sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N2O7S · xNa+ · yH2O
CAS Number:
Molecular Weight:
434.46 (anhydrous free acid basis)
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

[S](=O)(=O)(Nc2c(cccc2)C(=O)NCC(=O)O)c1ccc(cc1)OC(=O)C(C)(C)C

InChI

1S/C20H22N2O7S/c1-20(2,3)19(26)29-13-8-10-14(11-9-13)30(27,28)22-16-7-5-4-6-15(16)18(25)21-12-17(23)24/h4-11,22H,12H2,1-3H3,(H,21,25)(H,23,24)

InChI key

BTGNGJJLZOIYID-UHFFFAOYSA-N

Application

Sivelestat sodium salt hydrate may be used to inhibit human neutrophil elastase-mediated cell signaling.

Biochem/physiol Actions

Sivelestat is a competitive human neutrophil elastase (HNE) inhibitor (IC50 = 44 nM, Ki = 0.2 μM). It also inhibits leukocyte elastase obtained from rabbit, rat, hamster and mouse (IC50 = 19 to 49 nM). However, it does not inhibit trypsin, thrombin, plasmin, plasma kallikrein, pancreas kallikrein, chymotrypsin and cathepsin G even at 100 μM. In in-vivo studies, it suppressed lung hemorrhage in hamster (ID50 = 82 μg/kg) by intratracheal administration and increase of skin capillary permeability in guinea pig (ID50 = 9.6 mg/kg) by intravenous administration, both of which were induced by human neutrophil elastase.
Sivelestat is a competitive human neutrophil elastase (HNE) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kasturi Ganguly et al.
Frontiers in immunology, 13, 930449-930449 (2022-07-26)
Surfactant protein D (SP-D), a pattern recognition molecule, is emerging as a potent anti-tumoural innate immune defense molecule in a range of cancers. Previously, SP-D expression was found to be significantly downregulated at the malignant sites of human prostate adenocarcinoma
Eliana Pivetta et al.
Clinical science (London, England : 1979), 130(14), 1221-1236 (2016-02-28)
Lymphatic vasculature critically depends on the connections of lymphatic endothelial cells with the extracellular matrix (ECM), which are mediated by anchoring filaments (AFs). The ECM protein EMILIN1 is a component of AFs and is involved in the regulation of lymphatic
Yufeng Wang et al.
Frontiers in immunology, 13, 841445-841445 (2022-04-29)
The rate of complete remission of acute promyelocytic leukemia (APL) is currently over 90% because of the use of all-trans retinoic acid (ATRA) with arsenic trioxide (ATO). However, hemorrhagic mortality has emerged as the most significant barrier to APL-induced remission.
Biswajit Khatua et al.
Journal of leukocyte biology, 91(4), 641-655 (2012-01-14)
PA is an opportunistic pathogen that is commonly associated with severe infection in immunocompromised hosts. Siglec-9 binds with Sias by cis interaction on the neutrophil surface, thereby reducing immunological activity. However, neutrophils bind with pathogens through trans interactions of siglec-9
Katja Herges et al.
Multiple sclerosis (Houndmills, Basingstoke, England), 18(4), 398-408 (2012-02-22)
The pathology of neuromyelitis optica (NMO), in contrast to multiple sclerosis, comprises granulocyte infiltrates along extensive lengths of spinal cord, as well as optic nerve. Furthermore, IFN-β treatment worsens NMO. We recently found that experimental autoimmune encephalomyelitis (EAE) induced with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service