Skip to Content
Merck
All Photos(2)

Key Documents

E3520

Sigma-Aldrich

Ebselen

powder, ≥98% (TLC)

Synonym(s):

2-Phenyl-1,2-benzisoselenazol-3(2H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9NOSe
CAS Number:
Molecular Weight:
274.18
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Product Name

Ebselen, cysteine modifier

Quality Level

Assay

≥98% (TLC)

form

powder

mp

176-182  °C

solubility

chloroform: 19.60-20.40 mg/mL, clear, yellow

storage temp.

2-8°C

SMILES string

O=C1N([Se]c2ccccc12)c3ccccc3

InChI

1S/C13H9NOSe/c15-13-11-8-4-5-9-12(11)16-14(13)10-6-2-1-3-7-10/h1-9H

InChI key

DYEFUKCXAQOFHX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ebselen is a sympathomimetic amine.

Application

Ebselen has been used as a:
  • voltage-dependent calcium channels (VDCCs) blocker
  • glutathione peroxidase mimetic to test its inhibitory effect on contraction-mediated deoxy glucose (2-DG) uptake in mouse

extensor digitorum longus (fast-twitch) muscle
  • glucocerebrosidase inhibitor
  • hepatitis C virus helicase inhibitor

Biochem/physiol Actions

An organo-selenium compound possessing antioxidant properties. Inhibits mammalian lipoxygenases in the absence of thiol groups, and glutathione S-transferase and papain via the interaction with cysteine residues. Also inhibits indoleamine 2,3-dioxygenase by covalently modifying a cysteine residue, the effect being reversible with dithiothreitol. Inhibits oxidation of low density lipoproteins (LDL).
Ebselen, a glutathione-peroxidase-mimic, elicits anti-inflammatory activity. It also has anti-atherosclerotic functionality. Ebselen displays antibacterial action on) multidrug-resistant Staphylococcus aureus (MRSA) infections and could be a potential therapeutic target for treating MRSA based skin infection. It is regarded as neuroprotective agent and elicits chemopreventive functionality in inflammation-associated carcinogenesis.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ebselen, a glutathione peroxidase mimetic seleno-organic compound, as a multifunctional antioxidant implication for inflammation-associated carcinogenesis
Nakamura Y, et al.
The Journal of Biological Chemistry, 277(4), 2687-2694 (2002)
Andrew C Terentis et al.
Biochemistry, 49(3), 591-600 (2009-12-17)
The heme enzyme indoleamine 2,3-dioxygenase (IDO) plays an important immune regulatory role by catalyzing the oxidative degradation of l-tryptophan. Here we show that the selenezal drug ebselen is a potent IDO inhibitor. Exposure of human macrophages to ebselen inhibited IDO
Selcuk Yatmaz et al.
American journal of respiratory cell and molecular biology, 48(1), 17-26 (2012-09-25)
Oxidative stress caused by excessive reactive oxygen species production is implicated in influenza A virus-induced lung disease. Glutathione peroxidase (GPx)-1 is an antioxidant enzyme that may protect lungs from such damage. The objective of this study was to determine if
Sourav Mukherjee et al.
ACS chemical biology, 9(10), 2393-2403 (2014-08-16)
The hepatitis C virus (HCV) nonstructural protein 3 (NS3) is both a protease, which cleaves viral and host proteins, and a helicase that separates nucleic acid strands, using ATP hydrolysis to fuel the reaction. Many antiviral drugs, and compounds in
N Noguchi et al.
Biochimica et biophysica acta, 1213(2), 176-182 (1994-07-14)
The oxidative modification of low density lipoprotein (LDL) is accepted to be an important early event of atherosclerosis, but it has not yet been well understood. The preventive effects of two antioxidants with different functions, ebselen and probucol, against the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service