Skip to Content
Merck
All Photos(3)

Documents

S6936

Sigma-Aldrich

Anti-Sodium Channel, Pan antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-SP19

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot (chemiluminescent): 1:200-1:600

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Voltage-gated sodium channels (VGSC) are membrane proteins that are usually present in excitable cells. These sodium channels regulate neurological functions such as the generation of action potentials in nerve cells. Alterations in neurological sodium ion channels have been associated with epilepsy, whereas mutations in cardiac sodium channels have been linked to long QT syndrome . Anti-Sodium Channel, Pan antibody is specific for the α subunit of VGSC in rats.

Immunogen

synthetic peptide corresponding to amino acids 1491-1508 of the α subunit of rat type I voltage-gated sodium channel (VGSC, SP19) (with additional C-terminal cysteine). The epitope corresponds to the sequence in the intracellular loop between the III and IV domains of the VGSC α subunit.This epitope is identical in vertebrates, and highly homologous in mollusks and insects.

Application

Anti-Sodium Channel, Pan antibody is suitable for use in immunohistochemistry (formalin-fixed, paraffin-embedded sections), immunoprecipitation and chemiluminescent western blot.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Immunohistochemistry (1 paper)

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megan Oliva et al.
Epilepsia, 53(11), 1849-1859 (2012-08-22)
Voltage-gated sodium channels (VGSCs) are integral membrane proteins. They are essential for normal neurologic function and are, currently, the most common recognized cause of genetic epilepsy. This review summarizes the neurobiology of VGSCs, their association with different epilepsy syndromes, and
G M Vincent et al.
Cardiology in review, 7(1), 44-55 (1999-06-01)
The inherited long QT syndrome is caused by mutations of at least 5 ion channel genes. Mutations of the cardiac sodium ion channel gene and 3 potassium channel genes have been identified to this time. A genetic locus on chromosome
Margarita Calvo et al.
eLife, 5, e12661-e12661 (2016-04-02)
Neuropathic pain following peripheral nerve injury is associated with hyperexcitability in damaged myelinated sensory axons, which begins to normalise over time. We investigated the composition and distribution of shaker-type-potassium channels (Kv1 channels) within the nodal complex of myelinated axons following
X Liu et al.
Respiratory physiology & neurobiology, 178(3), 362-369 (2011-03-15)
Experiments in recent years have revealed labile electrophysiological and neurochemical phenotypes in primary afferent neurons exposed to specific stimulus conditions associated with the development of chronic pain. These studies collectively demonstrate that the mechanisms responsible for functional plasticity are primarily
Chunling Chen et al.
The Journal of biological chemistry, 287(46), 39061-39069 (2012-09-21)
Voltage-gated Na(+) channels in the brain are composed of a single pore-forming α subunit, one non-covalently linked β subunit (β1 or β3), and one disulfide-linked β subunit (β2 or β4). The final step in Na(+) channel biosynthesis in central neurons

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service