Skip to Content
Merck
All Photos(1)

Key Documents

AB144

Sigma-Aldrich

Anti-Choline Acetyltransferase Antibody

CHEMICON®, goat polyclonal

Synonym(s):

ChAT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Choline Acetyltransferase Antibody, serum, Chemicon®

biological source

goat

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

guinea pig, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Chat(12647)
rat ... Chat(290567)

General description

Acetylcholine (ACh) is a common neurotransmitter for motoneurons, preganglionic autonomic neurons, postganglionic parasympathetic neurons, a variety of brain regions and some emerging neuron-like stem cells. The metabolism of Ach is relatively simple, involving only two enzymes: choline acetyltransferase (ChAT) for synthesis and acetylcholinesterase (AChE) for degradation. Further, acetylcholine has little function in neurons other than neurotransmission and seems to be neuron specific. It seems that only cholinergic neurons have significant amounts of ChAT making anti-cholinacetyltransferase a useful specific marker.

Specificity

Recognizes cholinergic neurons in brain and central nervous systems.

Immunogen

Human placental enzyme.

Application

Anti-Choline Acetyltransferase Antibody is an antibody against Choline Acetyltransferase for use in IH & WB.
Immunoblot: 1:1,000-1:5,000

Immunohistochemistry: 1:500-1:1,000. See page two of this data sheet.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neuronal & Glial Markers

Target description

70 kDa

Physical form

Goat antisera, liquid, no preservatives.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Rat brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aging-related deficits in orexin/hypocretin modulation of the septohippocampal cholinergic system.
Stanley, EM; Fadel, J
Synapse null
Melissa A Stouffer et al.
Nature communications, 6, 8543-8543 (2015-10-28)
Insulin activates insulin receptors (InsRs) in the hypothalamus to signal satiety after a meal. However, the rising incidence of obesity, which results in chronically elevated insulin levels, implies that insulin may also act in brain centres that regulate motivation and
Fumiyasu Imai et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(21), 5724-5735 (2016-05-27)
Spinal reflex circuit development requires the precise regulation of axon trajectories, synaptic specificity, and synapse formation. Of these three crucial steps, the molecular mechanisms underlying synapse formation between group Ia proprioceptive sensory neurons and motor neurons is the least understood.
Lorelei Stoica et al.
Annals of neurology, 79(4), 687-700 (2016-02-19)
Amyotrophic lateral sclerosis (ALS) is a fatal neurodegenerative disease characterized by loss of motor neurons, resulting in progressive muscle weakness, paralysis, and death within 5 years of diagnosis. About 10% of cases are inherited, of which 20% are due to
Sally I Firth et al.
Experimental eye research, 75(1), 49-60 (2002-07-19)
The neuropeptide galanin has not been localized previously in the primate uvea, and the neuropeptide somatostatin has not been localized in the uvea of any mammal. Here, the distribution of galanin-like and somatostatin-like immunoreactive axons in the iris, ciliary body

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service