P5556
Poloxamer 188 solution
10%, sterile-filtered, BioReagent, suitable for insect cell culture
Synonym(s):
Nonionic copolymer surfactant
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12161902
PubChem Substance ID:
NACRES:
NA.75
Recommended Products
description
non-ionic
sterility
sterile-filtered
product line
BioReagent
mol wt
average mol wt 8350
concentration
10%
technique(s)
cell culture | insect: suitable
impurities
endotoxin, tested
transition temp
cloud point ≥100
HLB
29
SMILES string
O2C(C2)C.O1CC1
InChI
1S/C3H6O.C2H4O/c1-3-2-4-3;1-2-3-1/h3H,2H2,1H3;1-2H2
InChI key
RVGRUAULSDPKGF-UHFFFAOYSA-N
General description
Poloxamer 188 (P188) is a non-ionic amphiphilic copolymer. It comprises a central chain of hydrophobic polyoxypropylene flanked at ends by hydrophilic polyoxyethylene.
Application
A nonionic copolymer surfactant
Biochem/physiol Actions
Poloxamer 188 (P188) elicits hemorheological, antithrombotic, anti-inflammatory, and cytoprotective activities. It restores and stabilizes membrane barrier function. P188 also inhibits lipid peroxidation, reduces surface tension and hydrophobic-based cellular adherence. P-188 exhibits low toxicity and may be used in oral drug delivery.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Dina Mourad Saleh et al.
Particle and fibre toxicology, 17(1), 48-48 (2020-10-16)
Multi-walled carbon nanotubes can be divided into two general subtypes: tangled and straight. MWCNT-N (60 nm in diameter) and MWCNT-7 (80-90 nm in diameter) are straight-type MWCNTs, and similarly to asbestos, both are carcinogenic to the lung and pleura when administered to
Daniel Helman et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 85(2), 162-168 (2013-06-15)
We developed a membrane bound reporter and selection molecule for sorting by fluorescence activated cell sorting (FACS) of cells producing a protein of interest. This molecule is composed of a transmembrane (TM) domain, fused on its extracellular end to a
Deshka S Foster et al.
Nature communications, 11(1), 4061-4061 (2020-08-15)
Adhesions are fibrotic scars that form between abdominal organs following surgery or infection, and may cause bowel obstruction, chronic pain, or infertility. Our understanding of adhesion biology is limited, which explains the paucity of anti-adhesion treatments. Here we present a
Kerwin Kwek Zeming et al.
Lab on a chip, 20(18), 3445-3460 (2020-08-15)
In vitro erythroid cultures from human hematopoietic stem cells produce immature red blood cells (RBCs) called reticulocytes, which are important for RBCs production, and are widely used in scientific studies of malaria pathology, hematological diseases and protein translation. However, in
Rahul P Bakshi et al.
Nature communications, 9(1), 315-315 (2018-01-24)
Chemoprophylaxis is currently the best available prevention from malaria, but its efficacy is compromised by non-adherence to medication. Here we develop a long-acting injectable formulation of atovaquone solid drug nanoparticles that confers long-lived prophylaxis against Plasmodium berghei ANKA malaria in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service