Skip to Content
Merck

Skip To

B3259

Bradykinin acetate salt

≥98% (HPLC), Ca2+-dependent nitric oxide synthetase activator, powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C50H73N15O11 · xC2H4O2
CAS Number:
Molecular Weight:
1060.21 (free base basis)
MDL number:
UNSPSC Code:
12352106
EC Number:
227-781-2
NACRES:
NA.32
Assay:
≥98% (HPLC)
Form:
powder
Solubility:
0.1 M acetic acid: 25 mg/mL, clear, colorless
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Bradykinin acetate salt, powder, ≥98% (HPLC)

Quality Level

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white

solubility

0.1 M acetic acid: 25 mg/mL, clear, colorless

UniProt accession no.

storage temp.

−20°C

SMILES string

N2([C@@H](CCC2)C(=O)NCC(=O)N[C@@H](Cc5ccccc5)C(=O)N[C@@H](CO)C(=O)N3[C@@H](CCC3)C(=O)N[C@@H](Cc4ccccc4)C(=O)N[C@@H](CCCN=C(N)N)C(=O)O)C(=O)[C@H]1N(CCC1)C(=O)[C@@H](N)CCCN=C(N)N

InChI

1S/C50H73N15O11/c51-32(16-7-21-56-49(52)53)45(72)65-25-11-20-39(65)47(74)64-24-9-18-37(64)43(70)58-28-40(67)59-34(26-30-12-3-1-4-13-30)41(68)62-36(29-66)46(73)63-23-10-19-38(63)44(71)61-35(27-31-14-5-2-6-15-31)42(69)60-33(48(75)76)17-8-22-57-50(54)55/h1-6,12-15,32-39,66H,7-11,16-29,51H2,(H,58,70)(H,59,67)(H,60,69)(H,61,71)(H,62,68)(H,75,76)(H4,52,53,56)(H4,54,55,57)/t32-,33-,34-,35-,36-,37-,38-,39-/m0/s1

InChI key

QXZGBUJJYSLZLT-FDISYFBBSA-N

Gene Information

Application

Bradykinin acetate salt has been used:
  • to stimulate the contraction of guinea pig ileum
  • as a contractile agent in fibroblasts
  • In the stimulation of endothelium of human subcutaneous artery
  • To evoke Ca2+ signals in neuroblastoma-glioma hybrid cell line, NG108-15

Biochem/physiol Actions

Activates pain receptors; induces smooth muscle contraction; activates Ca2+-dependent nitric oxide synthetase; increases capillary permeability.
Bradykinin is a pro-inflammatory peptide that acts through G-protein-coupled receptors. It activates sphingolipid metabolism. Fibroblasts treated with bradykinin produce a rapid and important increase in ceramide which is followed by a transient rise in sphingosine content. The peptide also increases ceramide cellular content by rapidly mobilizing neutral glycolipids. Bradykinin plays an important role in the regulation of fluid and electrolyte balance, smooth muscle contraction, vasodilation and capillary permeability. The bradykinin potentiators, which possess no bradykinin activity themselves, enhance the activity of bradykinin.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
H4884S8758H7278
form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98%

assay

≥98% (TLC)

solubility

0.1 M acetic acid: 25 mg/mL, clear, colorless

solubility

acetic acid: 50 mg/mL, clear, colorless to very faintly yellow

solubility

50% acetic acid: 1 mg/mL, clear, colorless

solubility

acetic acid: 10 mg/mL, clear, colorless to faintly yellow

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

Gene Information

human ... KNG1 (includes EG:3827)(3827)

Gene Information

human ... ACE(1636), ACE2(59272)
mouse ... ACE3(217246)
rat ... ACE(11421), ACE2(70008), ACE3(498012)

Gene Information

-

Gene Information

-

storage condition

desiccated

storage condition

-

storage condition

-

storage condition

-


Still not finding the right product?

Explore all of our products under Bradykinin acetate salt


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Vasomotor dysfunction in human subcutaneous arteries exposed ex vivo to food-grade titanium dioxide
Jensen DM, et al.
Food And Chemical Toxicology, 120, 321-327 (2018)
Louis Potier et al.
The Journal of pharmacology and experimental therapeutics, 346(1), 23-30 (2013-04-18)
Cardiac ischemia is a leading cause of death, especially in diabetic patients. The diabetic ischemic heart is resistant experimentally to established cardioprotective treatments. New pharmacological approaches to cardiac protection are warranted. The kallikrein-kinin system is involved in myocardial protection in
C L Boyajian et al.
The Journal of biological chemistry, 266(8), 4995-5003 (1991-03-15)
The modulation of neuronal adenylylcyclase by Ca2+, acting via calmodulin, is a long-established example of a positive interaction between the Ca2(+)-mobilizing and cAMP-generating systems. In the present study, concentrations of Ca2+ that stimulate brain adenylylcyclase inhibit the adenylylcyclase of NCB-20



Global Trade Item Number

SKUGTIN
B3259-25MG04061833427514
B3259-10MG04061833427491
B3259-1MG04061833427507
B3259-5MG04061833427521
B3259-100MG04061833427484

Questions

  1. How should a solution of bradykinin acetate salt in 0.1M acetic acid be stored? Can it be further diluted in an aqueous buffer?

    1 answer
    1. Following reconstitution at 25 mg/mL in 0.1 M Acetic acid, aliquot and freeze at -20°C. Stock solutions are stable for up to 3 months at -20°C. The stability of dilute solutions has not been determined, however, Bradykinin fragment 1 - 7, (B4181) is part of a set of standards for Maldi-MS calibration (MSCAL1). In this application, the solution is diluted to 10 pmol/uL and stored at -20°C for 1 month. One may infer that dilute solutions of the whole molecule would be comparably stable when diluted and stored frozen.

      Helpful?

Reviews

No rating value

Active Filters