Skip to Content
Merck
All Photos(2)

Key Documents

S500HR

Sigma-Aldrich

Sephacryl® Size Exclusion Resin

500-HR, MW range 40-20,000 kDa (dextrans)

Synonym(s):

Poly([allyl dextran]-co-N,N′-methylenebisacrylamide)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

Product Name

Sephacryl®, 500-HR, MW range 40-20,000 kDa (dextrans)

Quality Level

description

stability: stable to 0.2M NaOH

form

suspension (20% aqueous ethanol)

feature

autoclavable

matrix

crosslinked allyl dextran/N,N′-methylenebisacrylamide copolymer

particle size

25-75 μm (wet)

pore size

40-20,000 kDa MW range (dextrans)

compatibility

mode of use size exclusion chromatography

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

  • Supplied swollen, suspended in 20% aqueous ethanol
  • Autoclavable
  • Stable to 0.2N sodium hydroxide (NaOH)
  • Store at refrigerated temperature (2-8 °C)
S500HR-750ML′s updated product number is GE17-0613-01

Application

For size exclusion chromatography; the HR grade is a smaller particle size with narrower size range distribution, allowing for more efficient separations at faster flow rates. Recommended fractionation ranges are listed for globular proteins and dextrans..
Sephacryl® has been used in size exclusion chromatography purification of the Acacia tortuosa gum, dextran-RBA12 from Weissella cibaria and dextran from Weissella cibaria JAG8.
Sephacryl® is used in gel filtration chromatography, protein chromatography, and resins and separation media. Sephacryl® has been used to develop a novel high-throughput screening assay for sickle cell disease drug discovery. Sephacryl® has also been used to purify and characterize ovine pancreatic elastase as well as a lysozyme from human urine.

Legal Information

Sephacryl is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 - 109.4 °F - closed cup

Flash Point(C)

38 - 43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Karymov et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 15(14), 2631-2641 (2001-12-01)
Dynamic alterations in chromatin structure mediated by postsynthetic histone modifications and DNA methylation constitute a major regulatory mechanism in DNA functioning. DNA methylation has been implicated in transcriptional silencing, in part by inducing chromatin condensation. To understand the methylation-dependent chromatin
Xiaolei Wang et al.
Molecular microbiology, 83(1), 195-207 (2011-11-25)
Stewartan and amylovoran exopolysaccharide (EPS) produced by the plant pathogenic bacteria Pantoea stewartii and Erwinia amylovora are virulence factors in the cause of Stewart's vascular wilt and fire blight. The biosynthesis of amylovoran and stewartan is encoded by a set
Structural and biocompatibility properties of dextran from Weissella cibaria JAG8 as food additive
Tingirikari JM, et al.
International Journal of Food Sciences and Nutrition, 65(6), 686-691 (2014)
Theresa L Gioannini et al.
The Journal of biological chemistry, 277(49), 47818-47825 (2002-10-10)
Experiments utilizing endotoxin aggregates, lipooligosaccharides (LOS) isolated from metabolically labeled Neisseria meningitidis serotype group B, demonstrate that albumin is an essential component of lipopolysaccharide binding protein- (LBP) and sCD14-dependent 1) disaggregation of LOS and 2) LOS activation of human umbilical
Nicole Iovine et al.
The Journal of biological chemistry, 277(10), 7970-7978 (2002-01-05)
The bactericidal/permeability increasing (BPI) and lipopolysaccharide (LPS)-binding (LBP) proteins are closely related two-domain proteins in which LPS binding is mediated by the NH(2)-terminal domain. To further define the role of the COOH-terminal domain of these proteins in delivery of LPS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service