Skip to Content
Merck
All Photos(1)

Documents

S400HR

Sigma-Aldrich

Sephacryl® Size Exclusion Resin

400-HR, MW range 20-8000 kDa (globular proteins)

Synonym(s):

Poly([allyl dextran]-co-N,N′-methylenebisacrylamide)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

product name

Sephacryl®, 400-HR, MW range 20-8000 kDa (globular proteins)

description

stability: stable to 0.2M NaOH

form

suspension (20% aqueous ethanol)

feature

autoclavable

matrix

crosslinked allyl dextran/N,N′-methylenebisacrylamide copolymer

particle size

25-75 μm (wet)

pore size

10-2000 kDa MW range (dextrans)
20-8000 kDa MW range (globular proteins)

compatibility

mode of use size exclusion chromatography

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

S400HR-750ML′s updated product is GE17-0609-01
  • Supplied swollen, suspended in 20% aqueous ethanol
  • Autoclavable
  • Stable to 0.2N sodium hydroxide (NaOH)
  • Store at refrigerated temperature (2-8 °C)

Application

Sephacryl® is used in gel filtration chromatography, protein chromatography, and resins and separation media. Sephacryl® has been used to develop a novel high-throughput screening assay for sickle cell disease drug discovery. Sephacryl® has also been used to purify and characterize enzymes such as bovine pancreatic elastase, tissue-nonspecific alkaline phosphatases and a lysozyme from human urine.
For size exclusion chromatography; the HR grade is a smaller particle size with narrower size range distribution, allowing for more efficient separations at faster flow rates. Recommended fractionation ranges are listed for globular proteins and dextrans..

Legal Information

Sephacryl is a registered trademark of Cytiva

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 - 109.4 °F - closed cup

Flash Point(C)

38 - 43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masaharu Tsuji et al.
International journal of systematic and evolutionary microbiology, 69(4), 944-948 (2019-01-04)
Four strains isolated from sediment sampled at the front of a retreating glacier on northern Ellesmere Island in the Canadian high Arctic, namely JCM 32575T, JCM 32576, JCM 32577 and JCM 32578, belong to a novel psychrophilic basidiomycetous yeast species
Michael J Lodes et al.
Methods in molecular medicine, 94, 91-106 (2004-02-13)
Expression cloning involves the selection of specific polypeptides, generated from a cDNA or genomic DNA library, based on certain characteristics of the expressed proteins, such as antibody or ligand binding, recognition by T-cells, function, or complementation of cell defects. Here
Sinu P John et al.
Journal of virology, 81(17), 8967-8976 (2007-06-15)
The Ebola virus (EBOV) genome encodes for several proteins that are necessary and sufficient for replication and transcription of the viral RNAs in vitro; NP, VP30, VP35, and L. VP30 acts in trans with an RNA secondary structure upstream of
Hypolipidemic and antioxidant activity of enoki mushrooms (Flammulina velutipes)
Ming-Yei Y, et al.
BioMed Research International (2014)
Bertrand Vileno et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(20), 8218-8223 (2011-05-04)
Double electron electron resonance EPR methods was used to measure the effects of the allosteric modulators, phosphorylation, and ATP, on the distances and distance distributions between the two regulatory light chain of myosin (RLC). Three different states of smooth muscle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service