Skip to Content
Merck
All Photos(2)

Documents

GE28-9356-06

Sigma-Aldrich

HiPrep 16/60 Sephacryl® S-500 HR

Cytiva 28-9356-06, column L × I.D. 60 cm × 16 mm, 50 μm avg. part. size

Synonym(s):

Sephacryl S-500 HR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

material

polypropylene (PP)

mol wt

40000-20000000 Da (globular proteins)

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 28-9356-06

parameter

<0.5 mL/min flow rate
<5 mL sample volume
0.15 Mpa (22 psi)

bed volume

120 mL

column L × I.D.

60 cm × 16 mm

matrix

dextran-acrylamide particle platform
allyl dextran and N, N’-methylenebisacrylamide
polymeric

matrix active group

dextran-acrylamide phase

avg. part. size

50 μm

cleaning

2-13

working range

3-11

separation technique

size exclusion (SEC)

General description

HiPrep 26/60 Sephacryl® S-500 HR are prepacked gel filtration column for preparative separation of biomolecules from up to 13 mL sample volume. Sephacryl® S-500 is used for separating polysaccarides, macromolecules with extended structures, and even small plasmids.

Features and Benefits

  • Maximum recovery and minimum nonspecific adsorption with long-term chemical and physical stability resulting from the hydrophilic, rigid allyl Dextran/bisacrylamide matrix.
  • Gel filtration columns for Low to medium pressure ÄKTA® systems.
  • Well-established in industrial production.
  • The HiPrep Sephacryl® series offers reproducible separations over a wide molecular weight range.
  • Convenient, prepacked HiPrep Sephacryl® columns speed up Protein purification.

Suitability

All commonly used buffers, 0.2 M NaOH, 0.1 M HCl, 1 M acetic acid, 8 M urea, 6 M guanidine HCl, 1 % SDS, 20 % ethanol, 30 % propanol, 30 % acetonitrile (tested at 40oC for 1 week), 0.5 M NaOH (only for cleaning-in-place)
gel filtration columns for low to medium pressure ÄKTA systems+AE14

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

HiPrep is a trademark of Cytiva
Sephacryl is a registered trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service