Skip to Content
Merck
All Photos(3)

Key Documents

S0327

Sigma-Aldrich

Scutellarein

≥98% (HPLC)

Synonym(s):

4′,5,6,7-Tetrahydroxyflavone, 6-Hydroxyapigenin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O6
CAS Number:
Molecular Weight:
286.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

biological source

Erigeron breviscapus

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

yellow

solubility

DMSO: 1 mg/mL, clear, greenish-yellow

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

Oc1ccc(cc1)C2=CC(=O)c3c(O)c(O)c(O)cc3O2

InChI

1S/C15H10O6/c16-8-3-1-7(2-4-8)11-5-9(17)13-12(21-11)6-10(18)14(19)15(13)20/h1-6,16,18-20H

InChI key

JVXZRQGOGOXCEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Flavonoid that is selectively cytotoxic toward cancer cells. Induces mitochondrial ROS, DNA damage, and metabolic suppression, preferentially in tumor cells. Scutellarein has been shown to have better absorption than scutellarin in Caco-2 monolayer cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenmiao Li et al.
Antiviral research, 177, 104714-104714 (2020-03-14)
Myricetin, a common dietary flavonoid, was reported to possess many different biological activities such as anti-oxidant, anti-inflammatory, and antiviral effects. In this study, we explored the anti-HSV effects and mechanisms of myricetin both in vitro and in vivo. The results
Mi-Sun Yu et al.
Bioorganic & medicinal chemistry letters, 22(12), 4049-4054 (2012-05-15)
Severe acute respiratory syndrome (SARS) is an infectious disease with a strong potential for transmission upon close personal contact and is caused by the SARS-coronavirus (CoV). However, there are no natural or synthetic compounds currently available that can inhibit SARS-CoV.
Xuesong Liu et al.
Journal of pharmaceutical sciences, 100(6), 2452-2459 (2011-01-20)
Antisolvent crystallization can be used as an alternative to cooling or evaporation for the separation and purification of solid product in the pharmaceutical industry. To improve the process understanding of antisolvent crystallization, the use of in-line tools is vital. In
Jian-feng Xing et al.
Acta pharmacologica Sinica, 32(5), 655-663 (2011-04-26)
To study the metabolic and pharmacokinetic profile of scutellarin, an active component from the medical plant Erigeron breviscapus (Vant) Hand-Mazz, and to investigate the mechanisms underlying the low bioavailability of scutellarin though oral or intravenous administration in rats. HPLC method
Xiuhua Hao et al.
Journal of pharmaceutical and biomedical analysis, 38(2), 360-363 (2005-06-01)
A validated HPLC method was developed for the quantification of scutellarin in rat plasma using a liquid-liquid extraction and an ultraviolet detection. Chromatographic separation of scutellarin in plasma was performed on a C18 column, with a mobile phase of acetonitrile-water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service