Skip to Content
Merck
All Photos(1)

Key Documents

SML0014

Sigma-Aldrich

Scutellarin hydrate

≥98% (HPLC)

Synonym(s):

5,6-Dihydroxy-2-(4-hydroxyphenyl)-4-oxo-4H-1-benzopyran-7-yl-ß-D-glucopyranosiduronic acid hydrate, 7-(β-D-Glucopyranuronosyloxy)-5,6-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one hydrate, Breviscapin, Scutellarein-7-O-β-D-glucuronide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18O12 · xH2O
CAS Number:
Molecular Weight:
462.36 (anhydrous basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

yellow

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1C(=O)O)O)O)O)Oc2cc3c(c(c2O)O)C(=O)C=C(O3)c4ccc(cc4)O

InChI

1S/C21H18O12/c22-8-3-1-7(2-4-8)10-5-9(23)13-11(31-10)6-12(14(24)15(13)25)32-21-18(28)16(26)17(27)19(33-21)20(29)30/h1-6,16-19,21-22,24-28H,(H,29,30)/t16-,17-,18+,19-,21+/m0/s1

InChI key

DJSISFGPUUYILV-ZFORQUDYSA-N

Application

Scutellarin hydrate may be used in MAPK-mediated cell signaling studies.

Biochem/physiol Actions

Scutellarin is a flavonoid that exhibits a number of cardio- and neuro-protective effects. Scutellarin inhibits p38 activity, ERK phosphorylation, and TGF-β expression and fibrosis in a rat myocardial infarct model. In a cerebral ischemia-reperfusion model, the compound reduced infarct volume and neurological deficits by increasing eNOS expression while inhibiting the induction of iNOS.

Features and Benefits

This compound is featured on the MAPKs and Nitric Oxide Synthases pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service