Skip to Content
Merck
All Photos(2)

Key Documents

RES6007H-A7

SAFC

HEPES sodium salt

Synonym(s):

4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17N2NaO4S
CAS Number:
Molecular Weight:
260.29
EC Number:
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.21

biological source

synthetic

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, microbial, and trace metals; tested

useful pH range

6.8-8.2

pKa (25 °C)

7.5

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

SMILES string

[Na+].OCCN1CCN(CC1)CCS([O-])(=O)=O

InChI

1S/C8H18N2O4S.Na/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14;/h11H,1-8H2,(H,12,13,14);/q;+1/p-1

InChI key

RDZTWEVXRGYCFV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Buffer quality is vital for the success of biopharmaceutical processes, because buffers are indispensable in nearly every production step.

Our broad portfolio of buffer materials manufactured under appropriate controls is tailored to your needs. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Application

HEPES Sodium is a biological buffer often referred to as a “Good′s” buffer. HEPES Sodium, used in conjunction with HEPES, is the most commonly used biological buffer formulation since the pKa of the buffer is 7.31 which corresponds to the physiological pH of 7.36. HEPES Sodium is considered to be non-toxic to culture cell lines, highly water soluble and provides high-solution clarity.

HEPES Sodium is used in cell culture media, biopharmaceutical buffer formulations (both upstream and downstream) and diagnostic reagent formulations.

Packaging

The product is available in the following package sizes:
RES6007H-A701X: 100g container
RES6007H-A702X: 1kg container
RES6007H-A704X: 10kg container
RES6007H-A705X: 25kg container

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anca G Delgado et al.
Microbial cell factories, 11, 128-128 (2012-09-15)
Buffering to achieve pH control is crucial for successful trichloroethene (TCE) anaerobic bioremediation. Bicarbonate (HCO3-) is the natural buffer in groundwater and the buffer of choice in the laboratory and at contaminated sites undergoing biological treatment with organohalide respiring microorganisms.
Peng Yang
Macromolecular bioscience, 12(8), 1053-1059 (2012-06-19)
When lysozyme is dissolved in a neutral HEPES buffer solution (pH = 7.4) with 0.001-0.050 M TCEP added, a fast phase transition process occurs and the resulting novel fiber-like hierarchical supramolecular assemblies made by primary spherical-particle aggregation can function as a "superglue"
Jihye Park et al.
The Analyst, 137(19), 4411-4414 (2012-08-07)
We have developed a quickly responsive, and specific fluorescent assay for the detection of Au(III) on the basis of the formation of gold nanoparticles in the presence of HEPES, which cleave the C-I bond of I-BODIPY 1 to yield the
J R Eidet et al.
Experimental eye research, 94(1), 109-116 (2011-12-14)
The use of amniotic membrane (AM) represents one of the major developments in ocular surface reconstruction. However, in a study on patients with primary pterygium, transplantation of AM with ex vivo expanded human conjunctival epithelial cells (HCjE) promoted earlier epithelialization
Tina Vognsen et al.
Biochemical and biophysical research communications, 420(1), 188-192 (2012-03-15)
The crystal structure of the NTF2-like domain of the Drosophila homolog of Ras GTPase SH3 Binding Protein (G3BP), Rasputin, was determined at 2.7Å resolution. The overall structure is highly similar to nuclear transport factor 2: It is a homodimer comprised

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service