RDD003
MOPS
≥99.5%, crystalline powder, anhydrous, Redi-Dri™
Synonym(s):
3-(N-Morpholino)propanesulfonic acid, 4-Morpholinepropanesulfonic acid
About This Item
Recommended Products
product name
MOPS, anhydrous, free-flowing, Redi-Dri™, ≥99.5%
grade
anhydrous
Quality Level
product line
Redi-Dri™
Assay
≥99.5%
form
crystalline powder
quality
free-flowing
pH
2.5-4 (25 °C, 209 g/L)
useful pH range
6.5-7.9
pKa (25 °C)
7.2
solubility
water: 0.5 g/mL, clear, colorless
λ
33 % in H2O
SMILES string
OS(=O)(=O)CCCN1CCOCC1
InChI
1S/C7H15NO4S/c9-13(10,11)7-1-2-8-3-5-12-6-4-8/h1-7H2,(H,9,10,11)
InChI key
DVLFYONBTKHTER-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- MOPS buffer for enhancing photoacoustic imaging: A study explored the utility of MOPS buffer in the synthesis of gold nanostars, enhancing their application in photoacoustic imaging due to the buffer′s pH stability, which is crucial for maintaining the optimal reaction conditions necessary for controlled nanostar growth (Zhang et al., 2024).
- MOPS for intracytoplasmic sperm injection (ICSI): MOPS buffer′s role was investigated in a study where its influx following membrane piercing during ICSI was found to alter the transcriptome of human oocytes, suggesting a significant impact on oocyte viability and fertilization outcomes (Mendola et al., 2024).
- Stability studies with MOPS in zinc complex formation: Utilizing MOPS buffer, a study examined the structures, solution equilibria, and speciation of zinc complexes with chloroquine and hydroxychloroquine, underlining the buffer′s role in maintaining the integrity and stability of these complexes (Squarcina et al., 2024).
- MOPS in radiolabeling and in vivo evaluation: MOPS buffer was integral in the radiolabeling processes of lanmodulin with biomedically relevant lanthanide isotopes, enhancing the precision and effectiveness of in vivo evaluations in biomedical research (Martin et al., 2023).
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service